Merck
  • Dipeptidase-1 Is an Adhesion Receptor for Neutrophil Recruitment in Lungs and Liver.
Dipeptidase-1 Is an Adhesion Receptor for Neutrophil Recruitment in Lungs and Liver.
Cell (2019-08-24)
Saurav Roy Choudhury, Liane Babes, Jennifer J Rahn, Bo-Young Ahn, Kimberly-Ann R Goring, Jennifer C King, Arthur Lau, Björn Petri, Xiaoguang Hao, Andrew K Chojnacki, Ajitha Thanabalasuriar, Erin F McAvoy, Sébastien Tabariès, Christoph Schraeder, Kamala D Patel, Peter M Siegel, Karen A Kopciuk, David C Schriemer, Daniel A Muruve, Margaret M Kelly, Bryan G Yipp, Paul Kubes, Stephen M Robbins, Donna L Senger
ABSTRACT
A hallmark feature of inflammation is the orchestrated recruitment of neutrophils from the bloodstream into inflamed tissue. Although selectins and integrins mediate recruitment in many tissues, they have a minimal role in the lungs and liver. Exploiting an unbiased in vivo functional screen, we identified a lung and liver homing peptide that functionally abrogates neutrophil recruitment to these organs. Using biochemical, genetic, and confocal intravital imaging approaches, we identified dipeptidase-1 (DPEP1) as the target and established its role as a physical adhesion receptor for neutrophil sequestration independent of its enzymatic activity. Importantly, genetic ablation or functional peptide blocking of DPEP1 significantly reduced neutrophil recruitment to the lungs and liver and provided improved survival in models of endotoxemia. Our data establish DPEP1 as a major adhesion receptor on the lung and liver endothelium and identify a therapeutic target for neutrophil-driven inflammatory diseases of the lungs.
Sigma-Aldrich
Anti-DPEP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Peroxidase from horseradish, lyophilized, powder, ~150 U/mg
Sigma-Aldrich
D-Amino Acid Oxidase from porcine kidney, powder, ≥1.5 units/mg solid
Sigma-Aldrich
Flavin adenine dinucleotide disodium salt hydrate, BioReagent, suitable for cell culture
Sigma-Aldrich
Methyl-β-cyclodextrin, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
4-Hydroxyphenylacetic acid, 98%
Sigma-Aldrich
Cilastatin sodium salt, ≥98% (HPLC)
Sigma-Aldrich
Trichrome Stain (Masson) Kit
Roche
In Situ Cell Death Detection Kit, Fluorescein, sufficient for ≤50 tests, suitable for detection