Merck
  • Fibroblast-Derived STC-1 Modulates Tumor-Associated Macrophages and Lung Adenocarcinoma Development.
Fibroblast-Derived STC-1 Modulates Tumor-Associated Macrophages and Lung Adenocarcinoma Development.
Cell reports (2020-06-25)
Tamihiro Kamata, Tsz Y So, Qasim Ahmed, Susan Giblett, Bipin Patel, Jinli Luo, Roger Reddel, Catrin Pritchard
ABSTRACT
The tumor microenvironment (TME) consists of different cell types, including tumor-associated macrophages (TAMs) and tumor-associated fibroblasts (TAFs). How these cells interact and contribute to lung carcinogenesis remains elusive. Using G12DKRAS- and V600EBRAF-driven mouse lung models, we identify the pleiotropic glycoprotein stanniocalcin-1 (STC1) as a regulator of TAM-TAF interactions. STC1 is secreted by TAFs and suppresses TAM differentiation, at least in part, by sequestering the binding of GRP94, an autocrine macrophage-differentiation-inducing factor, to its cognate scavenger receptors. The accumulation of mature TAMs in the Stc1-deficient lung leads to enhanced secretion of TGF-β1 and, thus, TAF accumulation in the TME. Consistent with the mouse data, in human lung adenocarcinoma, STC1 expression is restricted to myofibroblasts, and a significant increase of naive macrophages is detected in STC1-high compared with STC1-low cases. This work increases our understanding of lung adenocarcinoma development and suggests new approaches for therapeutic targeting of the TME.
Sigma-Aldrich
Rhein, technical grade
Sigma-Aldrich
GenElute Mammalian Total RNA Miniprep Kit, sufficient for 350 purifications
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-PDGF-AA Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone B-5-1-2, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Vimentin antibody produced in mouse, clone LN-6, ascites fluid