Merck
4D Genome Rewiring during Oncogene-Induced and Replicative Senescence.
Molecular cell (2020-03-30)
Satish Sati, Boyan Bonev, Quentin Szabo, Daniel Jost, Paul Bensadoun, Francois Serra, Vincent Loubiere, Giorgio Lucio Papadopoulos, Juan-Carlos Rivera-Mulia, Lauriane Fritsch, Pauline Bouret, David Castillo, Josep Ll Gelpi, Modesto Orozco, Cedric Vaillant, Franck Pellestor, Frederic Bantignies, Marc A Marti-Renom, David M Gilbert, Jean-Marc Lemaitre, Giacomo Cavalli
ABSTRACT
To understand the role of the extensive senescence-associated 3D genome reorganization, we generated genome-wide chromatin interaction maps, epigenome, replication-timing, whole-genome bisulfite sequencing, and gene expression profiles from cells entering replicative senescence (RS) or upon oncogene-induced senescence (OIS). We identify senescence-associated heterochromatin domains (SAHDs). Differential intra- versus inter-SAHD interactions lead to the formation of senescence-associated heterochromatin foci (SAHFs) in OIS but not in RS. This OIS-specific configuration brings active genes located in genomic regions adjacent to SAHDs in close spatial proximity and favors their expression. We also identify DNMT1 as a factor that induces SAHFs by promoting HMGA2 expression. Upon DNMT1 depletion, OIS cells transition to a 3D genome conformation akin to that of cells in replicative senescence. These data show how multi-omics and imaging can identify critical features of RS and OIS and discover determinants of acute senescence and SAHF formation.
Sigma-Aldrich
MISSION® esiRNA, targeting human GAPDH
Sigma-Aldrich
MISSION® esiRNA, targeting human KMT5C
Sigma-Aldrich
MISSION® esiRNA, targeting human DNMT3B
Sigma-Aldrich
MISSION® esiRNA, targeting human KMT5B
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys4) Antibody, clone MC315, rabbit monoclonal, culture supernatant, clone MC315, Upstate®
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer
Sigma-Aldrich
MISSION® esiRNA, targeting human RIF1
Sigma-Aldrich
MISSION® esiRNA, targeting human NTF4
Sigma-Aldrich
MISSION® esiRNA, targeting human LRWD1
Sigma-Aldrich
MISSION® esiRNA, targeting human DNMT3A
Sigma-Aldrich
MISSION® esiRNA, targeting human HMGA1
Sigma-Aldrich
MISSION® esiRNA, targeting human DNMT1