Merck
Integrin expression in cells of the intervertebral disc.
Journal of anatomy (2004-06-17)
Dana L Nettles, William J Richardson, Lori A Setton
ABSTRACT
In this study, we investigated the profile of integrin expression in human and porcine intervertebral disc tissue. Differences in extracellular matrix composition between anulus fibrosus (AF) and nucleus pulposus (NP) regions of the disc, as well as differences in cellular responses to environmental stimuli, suggest a role for integrins in presenting matrix signals that may mediate these responses. Human disc tissue and porcine AF and NP tissue were stained with antibodies to alpha integrin subunits 1-6, V and IIb, and beta integrin subunits 1-6 and graded for evidence of positive staining on a scale from 0 (no staining) to 3 (high incidence of staining). Human tissue expressed alpha and beta integrin subunits shown to be present in articular cartilage, including alpha(1), alpha(5) and alpha(V). Porcine AF tissue expressed similar integrin subunits to human disc, with both expressing alpha(1), alpha(5), beta(1), beta(3) and beta(5) subunits, whereas porcine NP tissue expressed higher levels of alpha(6), beta(1) and beta(4) than AF tissue. The expressed subunits are known to interact with proteins including collagens, fibronectin and laminin; however, additional studies will be required to characterize the interactions of the integrin subunits with specific matrix constituents, as well as their specific involvement in regulating environmental stimuli.
Sigma-Aldrich
Anti-Integrin α2 Antibody, clone P1E6, azide free, clone P1E6, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin α4 Antibody, clone P1H4, azide free, clone P1H4, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin β5 Antibody, CT/cytosolic, serum, Chemicon®
Sigma-Aldrich
Anti-Integrin α5 Antibody, CT, Intracellular, serum, Chemicon®
Sigma-Aldrich
Anti-Integrin α3 Antibody, clone P1B5, azide free, clone P1B5, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin β4 Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-Integrin β6 Antibody, clone CSβ6, azide free, clone CSbeta6, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin α6 Antibody, clone NKI-GoH3, clone NKI-GoH3, Chemicon®, from rat