Skip to Content
Merck
All Photos(4)

Key Documents

AB1928

Sigma-Aldrich

Anti-Integrin alpha 5 (ITGA5/CD49E) Antibody

CHEMICON®, rabbit polyclonal

Synonym(s):

CD49e, Fibronectin Receptor alpha Subunit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-Integrin α5 Antibody, CT, Intracellular, serum, Chemicon®

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, chicken, hamster, rat, mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
radioimmunoassay: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... ITGA5(3678)

General description

Integrins are a family of dimeric, transmembrane proteins that mediate cell-cell and extracellular matrix adhesion. Signals transduced by integrins play a role in many biological processes, including cell growth, differentiation, migration and apoptosis. The integrin family is composed of at least 15 alpha and 8 beta subunits that may form over twenty different alpha-beta non-covalently bound dimeric combinations on the cell surface. The alpha subunits all have some homology to each other, as do the beta subunits. Both of the subunits contribute to the binding of the ligand. Integrin alpha subunits contain seven weak sequence repeats in the N-terminal region, which may be important in ligand binding, and have been predicted to fold cooperatively into a single beta-propeller domain with seven beta-sheets. The alpha-3 subunit (CD49c) is highly concentrated in epithelial cells where it strongly adheres to Laminin-5 and Laminin-5 induced rapid adhesion can be blocked by antibodies against the alpha-3 integrin subunit. The alpha-3 subunit exists in two different splice variants, denoted as "A" and "B". The only difference that results from this differential splicing is a total change in the cytoplasmic domain, while the extracellular domain stays the same. Knock-out mice lacking this subunit show prenatal lethality and abnormalities in the kidneys.

Specificity

Antibody recognizes integrin alpha5. Specificity determined by immunoprecipitation from a detergent extract of 35S methionine labeled endothelial cells. No cross-reactivity to alpha1, alpha2, alpha3, alpha4, alpha6 or alphaV. The antibody recognizes an epitope on the cytoplasmic domain of the protein, and it is thus, intracellular.
Reactivity with other species has not been determined.

Immunogen

Synthetic peptide derived from the C-terminal sequence (cytoplasmic domain) of the human alpha5 integrin subunit.

Application

Anti-Integrin α5 Antibody, C-terminus, Intracellular detects level of Integrin α5 & has been published & validated for use in ELISA, IC, IF, IH, IP, RIA & WB.
Western Blot Analysis:
1:1000 dilution of a previous lot was used.

ELISA:
1:500-1:2000 dilution of a previous lot was used in ELISA.

Immunoprecipitation:
Recommended use of 5 μL of antibody for 5x106 cells.

Immunohistochemistry:
1:1000 dilution of a previous lot was used in tissue staining; suggested for use on acetone fixed tissue only.

Immunocytochemistry:
1:1000 of a previous lot was used in immunocytochemistry.

Optimal working dilutions must be determined by the end user.

Quality

Routinely evaluated by Western Blot on HUVEC lysates.

Western Blot Analysis: 1:1000 dilution of this lot detected integrin alpha 5 on 10 μg of HUVEC lysates.

Target description

114 kDa

Physical form

Rabbit polyclonal antiserum in buffer containing 0.05% sodium azide.

Analysis Note

Control
Mouse 3T3 fibroblasts Skin (Basement Membrane).

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mary Ann Stepp et al.
Molecular carcinogenesis, 49(4), 363-373 (2010-01-19)
Syndecan-1 (sdc-1) is a cell surface proteoglycan that mediates the interaction of cells with their matrix, influencing attachment, migration, and response to growth factors. In keratinocytes, loss of sdc-1 delays wound healing, reduces migration, and increases Transforming growth factor beta
S Khan et al.
Oncogene, 36(4), 491-500 (2016-06-21)
Although MUC13, a transmembrane mucin, is aberrantly expressed in pancreatic ductal adenocarcinoma (PDAC) and generally correlates with increased expression of HER2, the underlying mechanism remains poorly understood. Herein, we found that MUC13 co-localizes and interacts with HER2 in PDAC cells
Katsuyuki Nakamura et al.
Aging cell, 13(1), 175-184 (2013-11-20)
Aging causes phenotypic changes in skeletal muscle progenitor cells (Skm-PCs), such as reduced myogenesis and increased adipogenesis due to alterations in their environment or niche. Secreted protein acidic and rich in cysteine (SPARC), which is secreted into the niche of
Erpan Ahat et al.
Molecular biology of the cell, 30(6), 766-777 (2019-01-17)
The Golgi apparatus is a membrane-bound organelle that serves as the center for trafficking and processing of proteins and lipids. To perform these functions, the Golgi forms a multilayer stacked structure held by GRASP55 and GRASP65 trans-oligomers and perhaps their
Collagen VI is a basement membrane component that regulates epithelial cell-fibronectin interactions.
Groulx, JF; Gagne, D; Benoit, YD; Martel, D; Basora, N; Beaulieu, JF
Matrix Biology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service