Merck
  • Assessment of the integrin alpha5beta1 antagonist JSM6427 in proliferative vitreoretinopathy using in vitro assays and a rabbit model of retinal detachment.
Assessment of the integrin alpha5beta1 antagonist JSM6427 in proliferative vitreoretinopathy using in vitro assays and a rabbit model of retinal detachment.
Investigative ophthalmology & visual science (2009-10-10)
Grit Zahn, Kristine Volk, Geoffrey P Lewis, Dörte Vossmeyer, Roland Stragies, Jeffrey S Heier, Paul E Daniel, Anthony P Adamis, Ethan A Chapin, Steven K Fisher, Frank G Holz, Karin U Löffler, Jochen Knolle
ABSTRACT
To explore the role of integrin alpha5beta1 in proliferative vitreoretinopathy (PVR) pathogenesis by evaluating the expression alpha5beta1 on ARPE-19 cells and patient proliferative membranes, quantifying the inhibitory effects of JSM6427 (a small molecule alpha5beta1 inhibitor) on ARPE-19 cell adhesion and migration, and assessing the therapeutic potential of JSM6427 in a rabbit retinal detachment model. Expression of alpha5beta1 was evaluated on activated ARPE-19 cells by flow cytometry and on PVR membranes by immunohistochemistry. ARPE-19 cells were used in fibronectin-dependent adhesion and migration assays with various concentrations of JSM6427; IC(50) was calculated. In the rabbit model, eyes were intravitreally injected with vehicle or JSM6427 on day 0 or 1 after retinal detachment; BrdU was administered intravitreally on day 3, and retinal tissues were harvested on day 3 (4 hours later) or 7. Retinal scarring, cellular proliferation, and inflammatory responses were quantified, and retinal morphology was analyzed in retinal sections. Activated ARPE-19 cells and PVR membranes expressed high levels of alpha5beta1; expression was low in control eyes. JSM6427 provided a dose-dependent blockade of ARPE-19 cell adhesion to fibronectin (IC(50), 7.1 +/- 2.5 microM) and inhibition of migration (IC(50), 6.0 +/- 4.5 microM). In the rabbit model, intravitreal injection of JSM6427 provided significant inhibition of proliferation of retinal cells (Müller cells, microglia, and macrophages) on days 3 and 7 after detachment and inhibition of inflammatory response and retinal scarring on day 7 after detachment. JSM6427 is a promising treatment for PVR, with data suggesting that inhibition of alpha5beta1-fibronectin interactions addresses multiple pathways involving retinal pigment epithelial, glial, and inflammatory cells.
Sigma-Aldrich
Anti-Integrin αVβ5 Antibody, clone P1F6, azide free, clone P1F6, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin α5 Antibody, CT, Intracellular, serum, Chemicon®
Sigma-Aldrich
Anti-Integrin β1 Antibody, clone P4G11, FITC conjugated, clone P4G11, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin α5 Antibody, serum, Chemicon®