Merck
  • SIRT3 deficiency exacerbates p53/Parkin‑mediated mitophagy inhibition and promotes mitochondrial dysfunction: Implication for aged hearts.
SIRT3 deficiency exacerbates p53/Parkin‑mediated mitophagy inhibition and promotes mitochondrial dysfunction: Implication for aged hearts.
International journal of molecular medicine (2018-03-14)
Yan Li, Ying Ma, Liqiang Song, Lu Yu, Le Zhang, Yingmei Zhang, Yuan Xing, Yue Yin, Heng Ma
ABSTRACT
Mitochondrial dynamics have critical roles in aging, and their impairment represents a prominent risk factor for myocardial dysfunction. Mitochondrial deacetylase sirtuin (SIRT)3 contributes greatly to the prevention of redox stress and cell aging. The present study explored the role of SIRT3 on myocardium aging. Western blot analysis demonstrated that SIRT3 expression levels were significantly lower in the myocardia of aged mice compared with young mice. Immunoprecipitation and western blot assays indicated that the activity of mitochondrial manganese superoxide dismutase (MnSOD) and peroxisome proliferator‑activated receptor γ coactivator (PGC)‑1α was reduced in the aged heart. To further explore the association between SIRT3 and myocardial senescence, SIRT3 heart‑specific knockout (SIRT3-/-) mice were used in the present study. The results revealed that obvious features of aging were present in the myocardium of SIRT3-/- mice, including mitochondrial protein dysfunction, enhanced oxidative stress, and energy metabolism dysfunction. SIRT3 deficiency impaired Parkin‑mediated mitophagy by increasing p53‑Parkin binding and blocking the mitochondrial translocation of Parkin in cardiomyocytes. Injection of autophagy agonist CCCP significantly increased the mitochondrial Parkin level in young wild‑type hearts but not in aged hearts; the effect was less pronounced in SIRT3-/- hearts. These data suggest that CCCP‑induced Parkin translocation was reduced in aged and SIRT3-/- hearts. CCCP‑induced mitochondrial clearance, which could be rescued by autophagy antagonist bafilomycin‑A1, was markedly weakened in aged and SIRT3-/- hearts vs. young hearts. SIRT3 deficiency exacerbated p53/Parkin‑mediated mitophagy inhibition and disrupted mitochondrial homeostasis, suggesting that loss of SIRT3 may increase the susceptibility of aged hearts to cardiac dysfunction. Therapeutic activation of SIRT3 and improved mitochondrial function may ameliorate the symptoms of cardiac aging.
Sigma-Aldrich
Anti-Acetyl (Lys 319) p53 Antibody, C-Terminal antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Acetylated Lysine antibody produced in rabbit, 1 mg/mL, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-NDUFA9 antibody produced in mouse, clone 3D7, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
Sigma-Aldrich
Anti-SIRT1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-COX4I1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-p16 INK antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Superoxide Dismutase (MnSOD) (DD-17) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Sirt3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution