
Vinblastine sulfate

Synonym(s):
Vinblastine sulfate salt, VLB, Vincaleukoblastine sulfate salt
Empirical Formula (Hill Notation):
C46H58N4O9 · H2SO4
CAS Number:
Molecular Weight:
909.05
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3