Skip to Content
Merck
All Photos(1)

Documents

1713004

USP

Vinblastine sulfate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Vinblastine sulfate salt, Vincaleukoblastine, Sulfate (1:1) (salt), VLB, Vincaleukoblastine sulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H58N4O9 · H2SO4
CAS Number:
Molecular Weight:
909.05
Beilstein:
3659812
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

vinblastine

manufacturer/tradename

USP

mp

267 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

OS(O)(=O)=O.[H][C@@]12CN(CCc3c([nH]c4ccccc34)[C@@](C1)(C(=O)OC)c5cc6c(cc5OC)N(C)[C@@]7([H])[C@](O)([C@H](OC(C)=O)[C@]8(CC)C=CCN9CC[C@]67[C@]89[H])C(=O)OC)C[C@](O)(CC)C2

InChI

1S/C46H58N4O9.H2O4S/c1-8-42(54)23-28-24-45(40(52)57-6,36-30(15-19-49(25-28)26-42)29-13-10-11-14-33(29)47-36)32-21-31-34(22-35(32)56-5)48(4)38-44(31)17-20-50-18-12-16-43(9-2,37(44)50)39(59-27(3)51)46(38,55)41(53)58-7;1-5(2,3)4/h10-14,16,21-22,28,37-39,47,54-55H,8-9,15,17-20,23-26H2,1-7H3;(H2,1,2,3,4)/t28-,37-,38+,39+,42-,43+,44+,45-,46-;/m0./s1

InChI key

KDQAABAKXDWYSZ-PNYVAJAMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Vinblastine sulfate USP reference standard for use in specified quality tests and assays.

Also used to prepare standard and system suitability solutions for the assay, according to the given below monographs of United States Pharmacopeia (USP):
  • Vinblastine Sulfate
  • Vincristine Sulfate
  • Vinblastine Sulfate for Injection
  • Vincristine Sulfate for Injection
  • Vincristine Sulfate Injection

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Muta. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vincristine Sulfate for Injection
United States Pharmacopeia, 47(2) (2022)
Vincristine Sulfate
United States Pharmacopeia, 45(4) (2020)
Vinblastine Sulfate for Injection
United States Pharmacopeia, 47(2) (2022)
Vinblastine Sulfate
United States Pharmacopeia, 45(4) (2020)
Vincristine Sulfate Injection
United States Pharmacopeia, 47(2) (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service