Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Rotor for Eppendorf
®
5804/5804R & 5810/5810R Centrifuges
Products
Product Category
rotors (21)
Manufacturer
Package Size
Technique
Filter Products
Showing 1-20 of 27
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
EP5820715006
Rotor for Eppendorf
®
5804/5804R & 5810/5810R Centrifuges,
FA-45-6-30, aerosol tight, to hold, 6 x 15/50 mL conical tubes, lid
Expand
View Pricing
EP5820740000
Rotor for Eppendorf
®
5804/5804R & 5810/5810R Centrifuges,
S-4-104, for 5810/5810R, aerosol-tight, with 4 x 750 mL round buckets
Expand
View Pricing
EP5820760001
Rotor for Eppendorf
®
5804/5804R & 5810/5810R Centrifuges,
FA-45-48-11, aerosol tight, to hold, 48 x 1.5/2.0 mL tubes, lid
Expand
View Pricing
RA2DWP-EP
Rotor for Eppendorf
®
5804/5804R & 5810/5810R Centrifuges,
A-2-DWP, with 2 buckets
Expand
View Pricing
EP5804746007
Rotor for Eppendorf
®
5804/5804R & 5810/5810R Centrifuges,
S-4-72, with 4 × 250 mL round buckets
Expand
View Pricing
EP5820754001
Rotor for Eppendorf
®
5804/5804R & 5810/5810R Centrifuges,
S-4-104, for 5810/5810R, aerosol-tight, with 4 x 750 mL plate buckets
Expand
View Pricing
RA44450-EP
Rotor for Eppendorf
®
5804/5804R & 5810/5810R Centrifuges,
A-4-44, aerosol-tight, with 4 buckets for 2×50 mL conical tubes
Expand
View Pricing
RFA453011-EP
Rotor for Eppendorf
®
5804/5804R & 5810/5810R Centrifuges,
FA-45-30-11, aerosol tight, to hold, 30 x 1.5/2.0 mL tubes, lid
Expand
View Pricing
RA4624-EP
Rotor for Eppendorf
®
5804/5804R & 5810/5810R Centrifuges,
A-4-62, for 5810/5810R, aerosol-tight, with 4 MTP buckets
Expand
View Pricing
EP5820755008
Rotor for Eppendorf
®
5804/5804R & 5810/5810R Centrifuges,
S-4-104, for 5810/5810R, aerosol-tight, without buckets
Expand
View Pricing
RA462250-EP
Rotor for Eppendorf
®
5804/5804R & 5810/5810R Centrifuges,
A-4-62, for 5810/5810R, aerosol-tight, with 4 × 250 mL rectangular buckets
Expand
View Pricing
RA4814-EP
Rotor for Eppendorf
®
5804/5804R & 5810/5810R Centrifuges,
A-4-81, for 5810/5810R, aerosol-tight, with 4 MTP/Flex buckets
Expand
View Pricing
EP5820765003
Rotor for Eppendorf
®
5804/5804R & 5810/5810R Centrifuges,
FA-45-20-17, aerosol tight, to hold, 20 x 5.0 mL Eppendorf tubes, lid
Expand
View Pricing
RA481WO-EP
Rotor for Eppendorf
®
5804/5804R & 5810/5810R Centrifuges,
A-4-81, for 5810/5810R, aerosol-tight, without buckets
Expand
View Pricing
RA444100-EP
Rotor for Eppendorf
®
5804/5804R & 5810/5810R Centrifuges,
A-4-44, aerosol-tight, with 4 × 100 mL rectangular buckets
Expand
View Pricing
EP5820771003
Rotor for Eppendorf
®
5804/5804R & 5810/5810R Centrifuges,
F-35-48-17, to hold, 24 x 15 mL conical tubes, with 24 steel sleeves and adapters
Expand
View Pricing
EP5820772000
Rotor for Eppendorf
®
5804/5804R & 5810/5810R Centrifuges,
F-35-48-17, to hold, 40 x 15 mL conical tubes, with 48 steel sleeves and adapters
Expand
View Pricing
EP022639903
Rotor for Eppendorf
®
5804/5804R & 5810/5810R Centrifuges,
FA-45-6-30, aerosol-tight, to hold, 6 x 15/50 mL conical tubes, lid, including software upgrade (required for some older models)
Expand
View Pricing
RF453011-EP
Rotor for Eppendorf
®
5804/5804R & 5810/5810R Centrifuges,
F-45-30-11, non aerosol tight, to hold, 30 x 1.5/2.0 mL reaction vessels, lid
Expand
View Pricing
RA481500-EP
Rotor for Eppendorf
®
5804/5804R & 5810/5810R Centrifuges,
A-4-81, for 5810/5810R, aerosol-tight, with 4 × 500 mL rectangular buckets
Expand
View Pricing
Page 1 of 2
Page 1 of 2