Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
epT.I.P.S.
®
standard
Products
Product Category
pipette tips (13)
Manufacturer
Length (mm)
Volume (ml)
Volume (µL)
Color
Package Size
Size
Sterility
Filter Products
Showing 1-13 of 13
Compare
Product Number
Description
Pricing
EP022492098
Eppendorf
®
epT.I.P.S.
®
standard,
volume 0.5-10 mL, L 165 mm, turquoise, colorless tip, pkg of 200 ea (2 bags × 100 tips)
Expand
View Pricing
EP0030000650
Eppendorf
®
epT.I.P.S.
®
standard,
volume × L 0.2-5.0 mL × 175 mm , L, violet, pack of 300 ea (3 bags × 100 tips)
Expand
View Pricing
EP022492080
Eppendorf
®
epT.I.P.S.
®
standard,
volume 0.1-5 mL, L 120 mm, violet, colorless tip, pkg of 500 ea (5 bags × 100 ea)
Expand
View Pricing
ETIPRQ10-EP
Eppendorf
®
epT.I.P.S.
®
standard,
volume × L 0.5-10 mL × 243 mm , L, turquoise, pack of 200 ea (2 bags × 100 tips)
Expand
View Pricing
EP022492055
Eppendorf
®
epT.I.P.S.
®
standard,
volume 50-1,000 μL, L 71 mm, blue, colorless tip, pkg of 1000 ea (2 bags × 500 ea)
Expand
View Pricing
EP022492039
Eppendorf
®
epT.I.P.S.
®
standard,
volume 2-200 μL, L 53 mm, yellow, colorless tip, pkg of 1000 ea (2 bags × 500 ea)
Expand
View Pricing
EP022492071
Eppendorf
®
epT.I.P.S.
®
standard,
volume 0.25-2.5 mL, L 115 mm, red, colorless tip, pkg of 500 ea (5 bags × 100 ea)
Expand
View Pricing
EP022492004
Eppendorf
®
epT.I.P.S.
®
standard,
volume 0.1-10 μL, L 34 mm, dark gray, colorless tip, pkg of 1000 ea (2 bags × 500 ea)
Expand
View Pricing
EP022492021
Eppendorf
®
epT.I.P.S.
®
standard,
volume 0.5-20 μL, L 46 mm, light gray, colorless tip, pkg of 1000 ea (2 bags × 500 ea)
Expand
View Pricing
Z640077
Eppendorf
®
epT.I.P.S.
®
standard,
volume range 0.1-20 μL
Expand
View Pricing
EP022492047
Eppendorf
®
epT.I.P.S.
®
standard,
volume 20-300 μL, L 55 mm, orange, colorless tip, pkg of 1000 ea (2 bags × 500 ea)
Expand
View Pricing
EP022492063
Eppendorf
®
epT.I.P.S.
®
standard,
volume 50-1,250 μL, L 76 mm, green, colorless tip, pkg of 1000 ea (4 bags × 250 ea)
Expand
View Pricing
EP022492012
Eppendorf
®
epT.I.P.S.
®
standard,
volume 0.1-20 μL, L 40 mm, medium gray, colorless tip, pkg of 1000 ea (2 bags × 500 ea)
Expand
View Pricing