
Dantron

Synonym(s):
1,8-Dihydroxyanthraquinone, Chrysazine, Danthron
Empirical Formula (Hill Notation):
C14H8O4
CAS Number:
Molecular Weight:
240.21
Products
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Markush Class
Markush Group
API Family
Showing 1-1 of 1