Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Adapter for Eppendorf
®
A-4-44 Rotor
Products
Product Category
adapters (15)
Manufacturer
Diameter (mm)
Height (mm)
Package Size
Technique
Filter Products
Showing 1-15 of 15
Compare
Product Number
Description
Pricing
ADAPA444415-EP
Adapter for Eppendorf
®
A-4-44 Rotor,
holds 4 x Eppendorf Tube
®
5 mL & 15 mL conical tubes, for use with 100 mL rectangular bucket, pack of 2 ea
Expand
View Pricing
ADAPA44450FA-EP
Adapter for Eppendorf
®
A-4-44 Rotor,
holds 1 x 50 mL Falcon
®
centrifuge tubes, for 100 mL rectangular buckets, pack of 2 ea
Expand
View Pricing
ADAPA44475-EP
Adapter for Eppendorf
®
A-4-44 Rotor,
holds 1 x 50 -75 mL round-bottom tube, for 100 mL rectangular buckets, pack of 2 ea
Expand
View Pricing
ADAPA4444715-EP
Adapter for Eppendorf
®
A-4-44 Rotor,
holds 7 x 3 - 15 mL round-bottom tubes, for use with 100 mL rectangular bucket, pack of 2 ea
Expand
View Pricing
ADAPA4444927-EP
Adapter for Eppendorf
®
A-4-44 Rotor,
holds 9 x 2.6- 7 mL round-bottom tubes, for use with 100 mL rectangular bucket, pack of 2 ea
Expand
View Pricing
ADAPA44450F-EP
Adapter for Eppendorf
®
A-4-44 Rotor,
holds 1 x 50 mL Falcon
®
centrifuge tube, for 100 mL rectangular buckets with aerosol-tight cover, pack of 2 ea
Expand
View Pricing
ADAPA444125F-EP
Adapter for Eppendorf
®
A-4-44 Rotor,
holds 12 x 1.2 - 5 mL Falcon
®
centrifuge tubes, for 100 mL rectangular buckets, pack of 2
Expand
View Pricing
ADAPA44480-EP
Adapter for Eppendorf
®
A-4-44 Rotor,
holds 1 x 80 - 120 mL tubes, for 100 mL rectangular buckets, pack of 2 ea
Expand
View Pricing
ADAPA444125R-EP
Adapter for Eppendorf
®
A-4-44 Rotor,
holds 14 x 1.2 - 5 mL round-bottom tubes, for 100 mL rectangular buckets, pack of 2
Expand
View Pricing
ADAPA444418-EP
Adapter for Eppendorf
®
A-4-44 Rotor,
holds 4 x 7 - 18 mL round-bottom tubes, for use with 100 mL rectangular bucket, pack of 2 ea
Expand
View Pricing
ADAPA444215-EP
Adapter for Eppendorf
®
A-4-44 Rotor,
holds 2 x 15 mL conical tube, for 100 mL rectangular buckets, pack of 2
Expand
View Pricing
ADAPA444230-EP
Adapter for Eppendorf
®
A-4-44 Rotor,
holds 2 x 18 - 30 mL round-bottom tubes, for use with 100 mL rectangular bucket, pack of 2
Expand
View Pricing
ADAPA44450S-EP
Adapter for Eppendorf
®
A-4-44 Rotor,
capacity, 1 x 50 mL conical tube with skirted bottom, for 100 mL rectangular buckets, pack of 8
Expand
View Pricing
ADAPA44450-EP
Adapter for Eppendorf
®
A-4-44 Rotor,
holds 1 x 30 - 50 mL round-bottom / conical skirted tube, for use with 100 mL rectangular bucket, pack of 2 ea
Expand
View Pricing
ADAPA4444617-EP
Adapter for Eppendorf
®
A-4-44 Rotor,
holds 6 x 7 - 17 mL round-bottom tubes, for use with 100 mL rectangular bucket, pack of 2 ea
Expand
View Pricing