Skip to Content
Merck
All Photos(1)

Documents

1598201

USP

Raloxifene hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Keoxifene hydrochloride, LY 156758, [6-Hydroxy-2-(4-hydroxyphenyl)benzo[b]thien-3-yl][4-[2-(1-piperidinyl)ethoxy]phenyl]methanone hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H27NO4S · HCl
CAS Number:
Molecular Weight:
510.04
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

raloxifene

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl[H].Oc1ccc(cc1)-c2sc3cc(O)ccc3c2C(=O)c4ccc(OCCN5CCCCC5)cc4

InChI

1S/C28H27NO4S.ClH/c30-21-8-4-20(5-9-21)28-26(24-13-10-22(31)18-25(24)34-28)27(32)19-6-11-23(12-7-19)33-17-16-29-14-2-1-3-15-29;/h4-13,18,30-31H,1-3,14-17H2;1H

InChI key

BKXVVCILCIUCLG-UHFFFAOYSA-N

Gene Information

human ... ESR2(2100)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Raloxifene hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Raloxifene Hydrochloride Tablets

Biochem/physiol Actions

Raloxifene is a selective estrogen receptor modulator (SERM); acts as an anti-estrogen in both breast and uterine tissue while being estrogenic in bone. May have efficacy against estrogen-sensitive cancers.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K R Snyder et al.
American journal of health-system pharmacy : AJHP : official journal of the American Society of Health-System Pharmacists, 57(18), 1669-1675 (2000-09-28)
The pharmacology, pharmacokinetics, clinical efficacy, adverse effects, and therapeutic role of raloxifene hydrochloride are reviewed. Raloxifene is a selective estrogen-receptor modulator (SERM) that has been approved for use in the prevention and treatment of osteoporosis in postmenopausal women. A SERM
Tuan Hiep Tran et al.
Journal of nanoscience and nanotechnology, 14(7), 4820-4831 (2014-04-25)
The main aim of this study was to improve the oral bioavailability of raloxifene (RXF), a selective estrogen receptor modulator, by incorporation into solid lipid nanoparticles (SLN). RXF-loaded SLN was prepared by homogenization-sonication technique and characterized through physicochemical, pharmacokinetic, and
[Raloxifene hydrochloride (Evista Tablet 60 mg) for postmenopausal osteoporosis: mode of action and clinical efficacy].
Hiroyuki Kobayashi et al.
Nihon yakurigaku zasshi. Folia pharmacologica Japonica, 125(1), 37-48 (2005-03-02)
Roberta Fontana et al.
Endocrinology, 155(6), 2213-2221 (2014-03-19)
Estrogens play an important role in the regulation of energy homeostasis in female mammals and a reduced ovarian function, due to natural aging or surgery, is associated with body weight increase and fat redistribution. This disruption of energy homeostasis may
Ibrahim A Abdelazim et al.
Archives of osteoporosis, 9, 189-189 (2014-07-20)
Osteoporosis is a major cause of mortality and morbidity worldwide. Decreased bone turnover markers and increased lumbar spine and total hip bone mineral density (BMD) in raloxifene-treated women add further support to the idea that raloxifene is an effective well-tolerated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service