Skip to Content
Merck
All Photos(1)

Documents

1551503

USP

Povidone

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Polyvinylpyrrolidone, PVP, Polyvidone, Povidone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H9NO)n
CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

povidone

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

C=CN1CCCC1=O

InChI

1S/C6H9NO/c1-2-7-5-3-4-6(7)8/h2H,1,3-5H2

InChI key

WHNWPMSKXPGLAX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Povidone USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Vigabatrin for Oral Solution
  • Vigabatrin Tablets

Other Notes

Polyvinylpyrrolidone is a component of Denhardt′s Solution and is included at a concentration of 1% (w/v) in the standard 50X stock solution.
Sales restrictions may apply.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rozhin Penjweini et al.
Biochimica et biophysica acta, 1843(5), 855-865 (2014-02-01)
PVP-Hypericin (PVP: polyvinylpyrrolidone) is a potent anti-cancer photosensitizer for photodynamic diagnosis (PDD) and therapy (PDT). However, cellular targets and mechanisms involved in the cancer-selectivity of the photosensitizer are not yet fully understood. This paper gives new insights into the differential
Atsuko Adachi et al.
Contact dermatitis, 48(3), 133-136 (2003-05-21)
A 59-year-old woman who had had several episodes of contact urticaria after hair treatment, developed anaphylaxis after vaginal application of povidone-iodine solution for disinfection. Prick tests showed wheal-and-flare responses to both povidone-iodine (0.1% aqueous) and polyvinylpyrrolidone (povidone, PVP) (0.001% aq.)
[Gordon's polyvinylpyrrolidone test].
J Aebersold
Helvetica medica acta, 32(3), 227-246 (1965-09-01)
Povidone-iodine solutions in traumatic wound preparation.
M S Oberg
The American journal of emergency medicine, 5(6), 553-555 (1987-11-01)
Patricia C Buchsel
Expert opinion on drug metabolism & toxicology, 4(11), 1449-1454 (2008-10-28)
Painful oral lesions in patients with cancer, HIV or aphthous ulcers can cause significant pain and discomfort. Those with oral mucositis (OM) resulting from high-dose chemotherapy or irradiation are the most at risk for painful lesions that lead to significant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service