Skip to Content
Merck
All Photos(1)

Key Documents

1448901

USP

Mupirocin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

5,9-Anhydro-2,3,4,8-tetradeoxy-8-[[3-(2-hydroxy-1-methylpropyl)oxiranyl]methyl]-3-methyl-[2E,8[2S,3S(1S,2S)]]-L-talonon-2-enonic acid 8-carboxyoctyl ester, BRL 4910A, Pseudomonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H44O9
CAS Number:
Molecular Weight:
500.62
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

mupirocin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H][C@@]1(CO[C@@H](C\C(C)=C\C(=O)OCCCCCCCCC(O)=O)[C@H](O)[C@@H]1O)C[C@@H]2O[C@@]2([H])[C@@H](C)[C@H](C)O

InChI

1S/C26H44O9/c1-16(13-23(30)33-11-9-7-5-4-6-8-10-22(28)29)12-20-25(32)24(31)19(15-34-20)14-21-26(35-21)17(2)18(3)27/h13,17-21,24-27,31-32H,4-12,14-15H2,1-3H3,(H,28,29)/b16-13+/t17-,18-,19-,20-,21-,24+,25-,26-/m0/s1

InChI key

MINDHVHHQZYEEK-HBBNESRFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Mupirocin USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Biochem/physiol Actions

Antibiotic; inhibits isoleucyl-tRNA synthetase (IRS).

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M E H Jaspers et al.
Burns : journal of the International Society for Burn Injuries, 40(8), 1570-1574 (2014-04-02)
Staphylococcus aureus wound colonization frequently occurs in patients with burns and can cause impaired wound healing. Nasal mupirocin application may contribute to the reduction of burn wound colonization of endogenous origin, whereas colonization by the exogenous route can be reduced
Miranda M L van Rijen et al.
The Journal of antimicrobial chemotherapy, 61(2), 254-261 (2008-01-05)
The majority of nosocomial Staphylococcus aureus infections originate from the patients' own flora, with nasal carriage of S. aureus before surgical procedures being a risk factor for subsequent infection. The objective of this review was to assess whether intranasal mupirocin
Christopher M Thomas et al.
Nature reviews. Microbiology, 8(4), 281-289 (2010-03-02)
Mupirocin, a polyketide antibiotic produced by Pseudomonas fluorescens, is used to control the carriage of methicillin-resistant Staphylococcus aureus on skin and in nasal passages as well as for various skin infections. Low-level resistance to the antibiotic arises by mutation of
Gaosi Xu et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 25(2), 587-592 (2009-08-15)
Recently, there have been increasing concerns about the emergence of mupirocin resistance and increased infections due to lowered inhibition of Staphylococcus aureus. We conducted this systemic analysis to find out whether the application of mupirocin was effective for the prevention
I Eltringham
The Journal of hospital infection, 35(1), 1-8 (1997-01-01)
Mupirocin has become the topical agent of choice for the elimination of methicillin-resistant Staphylococcus aureus (MRSA) carriage. The increased use of this antibiotic has been followed by reports of outbreaks due to MRSA with both low- and high-level resistance. Whilst

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service