Skip to Content
Merck
All Photos(1)

Key Documents

1379401

USP

Meloxicam

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

4-Hydroxy-2-methyl-N-(5-methyl-2-thiazolyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H13N3O4S2
CAS Number:
Molecular Weight:
351.40
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

meloxicam

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

InChI

1S/C14H13N3O4S2/c1-8-7-15-14(22-8)16-13(19)11-12(18)9-5-3-4-6-10(9)23(20,21)17(11)2/h3-7,19H,1-2H3,(H,15,16)/b13-11+

InChI key

DWMREKMVXIFPFM-ACCUITESSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Meloxicam USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Meloxicam Oral Suspension
  • Meloxicam Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Türck et al.
British journal of rheumatology, 35 Suppl 1, 13-16 (1996-04-01)
Meloxicam is a new preferential cyclooxygenase-2 (COX-2) inhibitor currently for the treatment of osteoarthritis and rheumatoid arthritis. Its pharmacokinetic profile is characterized by a prolonged and almost complete absorption and the drug is > 99.5% bound to plasma proteins. Meloxicam
[Meloxicam (Movalis)].
Dusanka Martinović Kaliterna
Reumatizam, 54(2), 81-82 (2008-03-21)
S Noble et al.
Drugs, 51(3), 424-430 (1996-03-01)
Meloxicam is a new nonsteroidal anti-inflammatory drug (NSAID) developed for the treatment of rheumatoid arthritis and osteoarthritis. It has greater in vitro and in vivo inhibitory action against the inducible isoform of cyclo-oxygenase (COX-2), which is implicated in the inflammatory
[The new nonsteroidal anti-inflammatory preparation meloxicam in the treatment of rheumatoid arthritis].
E I Grishina
Likars'ka sprava, (2)(2), 19-25 (1997-03-01)
Mansoor Ahmed et al.
Expert opinion on drug metabolism & toxicology, 1(4), 739-751 (2006-07-26)
Rheumatoid arthritis (RA), a chronic inflammatory multisystem disorder marked by joint pain, stiffness, swelling and multiple systemic involvements, requires a multifaceted approach for its management. It includes symptomatic treatment with analgesics as well as disease-modifying medications to alter the course

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service