Skip to Content
Merck
All Photos(1)

Key Documents

1349626

USP

Isomalt

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

6-O-α-D-Glucopyranosyl-D-glucitol mixed with 1-O-α-D-glucopyranosyl-D-mannitol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H24O11
CAS Number:
Molecular Weight:
344.31
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

isomalt

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O1[C@@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)OC[C@@H](O)[C@@H](O)[C@H](O)C(O)CO

InChI

1S/C12H24O11/c13-1-4(15)7(17)8(18)5(16)3-22-12-11(21)10(20)9(19)6(2-14)23-12/h4-21H,1-3H2/t4?,5-,6-,7-,8-,9-,10+,11-,12+/m1/s1

InChI key

SERLAGPUMNYUCK-BLEZHGCXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Isomalt USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Holub et al.
Hormone and metabolic research = Hormon- und Stoffwechselforschung = Hormones et metabolisme, 41(12), 886-892 (2009-08-25)
The polyol isomalt (Palatinit) is a very low glycaemic sugar replacer. The effect of food supplemented with isomalt instead of higher glycaemic ingredients like sucrose and/or starch hydrolysates on metabolic control in patients with type 2 diabetes was examined in
A D'Alessandro et al.
La Clinica terapeutica, 150(1), 29-32 (1999-06-15)
This study investigates the bowel function in outdoor runners drinking sweetened solutions both by determination of H2 in exhaled air and asking about intestinal discomfort. Twenty-two athletes with a mean age of 22 +/- 3 yrs, received 100 ml of
A Gostner et al.
The British journal of nutrition, 94(4), 575-581 (2005-10-04)
The polyol isomalt (Palatinit) is a well established sugar replacer. The impact of regular isomalt consumption on metabolism and parameters of gut function in nineteen healthy volunteers was examined in a randomised, double-blind, cross-over trial with two 4-week test periods.
Tsutomu Takatsuka et al.
Clinical oral investigations, 12(2), 173-177 (2007-12-25)
Isomalt is a non-cariogenic sweetener, which is widely used in sugar-free candy and chewing gum. Little is known about the effects of Isomalt on de- and remineralization. Binding between calcium and Isomalt has been reported, which could affect the mineral
F Ndindayino et al.
International journal of pharmaceutics, 235(1-2), 149-157 (2002-03-07)
Isomalt, a sugar alcohol, was melt-extruded prior to compression in order to improve its tabletting properties. After fusion, crystalline isomalt was transformed into an amorphous form as shown by X-ray diffraction and differential scanning calorimetry (DSC). The tabletting properties of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service