Skip to Content
Merck
All Photos(1)

Key Documents

1233009

USP

Edetate disodium

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Ethylenediaminetetraacetic acid disodium salt dihydrate, Disodium ethylenediaminetetraacetate dihydrate, EDTA disodium salt, EDTA-Na2, Edathamil, Edetate disodium salt dihydrate, Sequestrene Na2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2Na2O8 · 2H2O
CAS Number:
Molecular Weight:
372.24
Beilstein:
3900609
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

edta

manufacturer/tradename

USP

mp

248 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O.O.O.O.[Na+].[Na+].[Na+].[Na+].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC(O)=O.OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

InChI

1S/C10H16N2O8.2Na.2H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;2*1H2/q;2*+1;;/p-2

InChI key

OVBJJZOQPCKUOR-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Chelator of divalent cations. Inhibits enzymes, such as metalloproteases, that require divalent cations for activity.
Edetate disodium USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Edetate Disodium Injection
  • Edetate Disodium Compounded Ophthalmic Solution
  • Levobunolol Hydrochloride Ophthalmic Solution
  • Budesonide Nasal Spray

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Levobunolol Hydrochloride Ophthalmic Solution
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 44(4), 2595-2595 (2019)
Edetate Disodium Compounded Ophthalmic Solution
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 44(4) (2021)
Il Joong Park et al.
Archives of pathology & laboratory medicine, 138(8), 1077-1082 (2014-07-31)
The Samsung LABGEO(HC10) Hematology Analyzer (LABGEO(HC10)) is a recently developed automated hematology analyzer that uses impedance technologies. The analyzer provides 18 parameters including 3-part differential at a maximum rate of 80 samples per hour. To evaluate the performance of the
David Voglar et al.
Environmental technology, 35(9-12), 1389-1400 (2014-04-08)
We demonstrate here, in a pilot-scale experiment, the feasibility of ethylenediaminetetraacetate (EDTA)based washing technology for soils contaminated with potentially toxic metals. Acid precipitation coupled to initial alkaline toxic metal removal and an electrochemical advanced oxidation process were used for average
Pseudothrombocytopenia in a patient receiving romiplostim for immune thrombocytopenic purpura.
Claudio Fozza et al.
Annals of hematology, 93(5), 899-900 (2013-10-02)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service