Skip to Content
Merck
All Photos(1)

Key Documents

1086141

USP

Calcipotriene

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Calcipotriol, (5Z,7E,22E,24S)-24-Cyclopropyl-9,10-secochola-5,7,10(19),22-tetraene-1α,3ε,24-triol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H40O3
CAS Number:
Molecular Weight:
412.60
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

calcipotriene

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O[C@H]1C[C@@H](C(=C)\C(=C/C=C2/C3[C@@]([C@H](CC3)[C@H](C)\C=C\[C@@H](O)C4CC4)(CCC/2)C)\C1)O

InChI

1S/C27H40O3/c1-17(6-13-25(29)20-8-9-20)23-11-12-24-19(5-4-14-27(23,24)3)7-10-21-15-22(28)16-26(30)18(21)2/h6-7,10,13,17,20,22-26,28-30H,2,4-5,8-9,11-12,14-16H2,1,3H3/b13-6+,19-7+,21-10-/t17-,22-,23-,24?,25-,26+,27-/m1/s1

InChI key

LWQQLNNNIPYSNX-OUSHRDKSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Calcipotriene exists in two forms, anhydrous, which does not contain water molecules, or hydrated, which contains one molecule of water.

Application

Calcipotriene USP reference standard for use in specified quality tests and assays.

Also used to prepare system suitability, standard, sensitivity solution, standard stock, and system suitability stock solution for assay and impurity analysis according to the given below monographs of United States Pharmacopeia (USP):
  • Calcipotriene
  • Calcipotriene Ointment
  • Calcipotriene Cream

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Oral - Eye Irrit. 2 - Repr. 2 - Skin Sens. 1B - STOT RE 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Calcipotriene
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 45(5) (2021)
Calcipotriene Cream
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 45(6) (2021)
Calcipotriene Ointment
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 45(5) (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service