Skip to Content
Merck
All Photos(1)

Key Documents

K08670356

Kromasil® C18 HPLC Column

5 μm particle size, pore size 100 Å, L × I.D. 150 mm × 4.6 mm

Synonym(s):

Kromasil C18

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52
Pricing and availability is not currently available.

Agency

suitable for USP L1

product line

Kromasil®

manufacturer/tradename

Kromasil® KR100-5-C18-4.6X150

technique(s)

HPLC: suitable

L × I.D.

150 mm × 4.6 mm

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

100 Å

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Reverse phase (RP) HPLC columns are majorly advantageous in ensuring high precision data, they are widely available commercially and are easy to use and less time-consuming.[1] Two main approaches one should consider before choosing a RP column are, firstly characterising physiochemical parameters of the packing material such as 1% carbon load, surface area, pore size, particle size and pH. These are some of the important parameters important to control the quality of the column. Second approach is based upon the chromatographic and statistical evaluation of the retention behaviour for a large number of compounds. Kromasil C18 columns are basically RP columns made up of silica-based monomeric type reversed-phase packing material.[2]

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Kromasil is a registered trademark of EKA Chemicals AB

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Kele et al.
Journal of chromatography. A, 855(2), 423-453 (1999-10-16)
The reproducibility of the retention data and the band profiles was investigated with Kromasil C18 columns (silica-based monomeric type reversed-phase packing material). High precision data were obtained and statistically compared among five columns from the same batch (column-to-column reproducibility) and
Chromatographic classification of commercially available reverse-phase HPLC columns.
Cruz E
Chromatographia, 44 (3-4), 151-161 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service