Skip to Content
Merck
All Photos(1)

Key Documents

CRM48617

Supelco

Benzene solution

certified reference material, TraceCERT®, 200 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6
CAS Number:
Molecular Weight:
78.11
MDL number:
UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

pkg of 1 mL

concentration

200 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-30°C

SMILES string

c1ccccc1

InChI

1S/C6H6/c1-2-4-6-5-3-1/h1-6H

InChI key

UHOVQNZJYSORNB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Letter to the editor in response to 'Low-dose metabolism of benzene in humans: science and obfuscation' Rappaport et al. (2013).
Paul S Price et al.
Carcinogenesis, 34(7), 1692-1696 (2013-03-28)
Angela M Moro et al.
Mutation research, 754(1-2), 63-70 (2013-05-01)
We evaluated genotoxic effects of exposure to low levels of benzene, a class I human carcinogen, among gasoline station attendants (GSA). Oxidative stress and the protective effects of antioxidants on DNA damage were also analyzed. Although exposures were below ACGIH
Authors' response to comments on Petty et al. (2011), "a quantitative method for estimating dermal benzene absorption from benzene containing hydrocarbon liquids," IJOEH, 17:287-300 by Pamela R.D. Williams, Jennifer Sahmel, Annette L.Bunge, Jeffrey Knutsen, and John Spencer.
Stephen E Petty et al.
International journal of occupational and environmental health, 19(2), 147-154 (2013-07-20)
Qingfen Niu et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 107, 377-385 (2013-03-15)
Three novel planar and star-shaped molecules containing thiophene-functionalized group and acetylenic spacers, 4-((5″-iodo-[2,2':5',2″-terthiophen]-5-yl)ethynyl)aniline (I3TEA), 4,4'-([2,2':5',2″-terthiophene]-5,5″-diylbis(ethyne-2,1-diyl))dianiline (3TDDA), 1,3,5-tris((5-((trimethylsilyl)ethynyl)thiophen-2-yl)ethynyl)benzene (TETEB) were synthesized through Sonogashira coupling reaction. Their photophysical and electrochemical properties were explored by UV-vis, photoluminescence (PL) emission and DFT calculation. Three
Wenliang Huang et al.
Nature communications, 4, 1448-1448 (2013-02-07)
Aromaticity is a fundamental concept with implications spanning all the chemical sciences. Hückel's (4n+2)π-electron rule is the standard criterion to determine aromaticity and it applies well to neutral arenes as well as to charged species such as the cyclopentadienyl anion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service