CRM46975
Volatile Free Acid Mix
certified reference material, bottle of 100 mL
Synonym(s):
VFA mix
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
85151701
NACRES:
NA.24
Recommended Products
grade
certified reference material
TraceCERT®
Quality Level
product line
TraceCERT®
CofA
current certificate can be downloaded
packaging
bottle of 100 mL
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
format
multi-component solution
storage temp.
2-8°C
Application
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Other Notes
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Legal Information
TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany
Analyte
Description
Acetic acid
Butyric acid
Formic acid
Heptanoic acid
Hexanoic acid
Isobutyric acid
Isovaleric acid
4-Methylvaleric acid
Propionic acid
Valeric acid
See All (10)
related product
Product No.
Description
Pricing
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
María J Trujillo-Rodríguez et al.
Talanta, 172, 86-94 (2017-06-13)
A vacuum headspace single-drop microextraction method based on the use of magnetic ionic liquids (vacuum MIL-HS-SDME) for the determination of short chain free fatty acids is described for the first time. The basis of the method involves the use of
Neil Yohan Musadji et al.
Molecules (Basel, Switzerland), 25(22) (2020-11-18)
Dissolved organic matter is an important component of the global carbon cycle that allows the distribution of carbon and nutrients. Therefore, analysis of soil dissolved organic matter helps us to better understand climate change impacts as it is the most
Iulian A Grosu et al.
Frontiers in veterinary science, 7, 31-31 (2020-03-13)
Microbiota affects host health and plays an important role in dysbiosis. The study examined the effect of diet including grape seed meal (GSM) with its mixture of bioactive compounds on the large intestine microbiota and short-chain fatty acid synthesis in
Lisa Elmén et al.
Frontiers in microbiology, 11, 892-892 (2020-06-06)
Dietary emulsifiers are widely used in industrially processed foods, although the effects of these food additives on human gut microbiota are not well studied. Here, we investigated the effects of five different emulsifiers [glycerol monoacetate, glycerol monostearate, glycerol monooleate, propylene
Alberto Amaretti et al.
International journal of food sciences and nutrition, 71(5), 563-571 (2020-01-09)
Dextrans are homopolysaccharides of D-glucose units produced by lactic acid bacteria. They have several technological applications and potential utilisation in positively modulating gut microbiota is attracting increasing attention. Whereas the prebiotic activity of low polymerisation degree (DP) dextrans has been
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service