Skip to Content
Merck
All Photos(1)

Key Documents

CRM03054

Supelco

Tetramethrin

mixture of stereoisomers, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H25NO4
CAS Number:
Molecular Weight:
331.41
Beilstein:
8807938
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

composition

cis-Tetramethrin, <25.0% (minor component)

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage temp.

−20°C

SMILES string

C\C(C)=C\C1C(C(=O)OCN2C(=O)C3=C(CCCC3)C2=O)C1(C)C

InChI

1S/C19H25NO4/c1-11(2)9-14-15(19(14,3)4)18(23)24-10-20-16(21)12-7-5-6-8-13(12)17(20)22/h9,14-15H,5-8,10H2,1-4H3

InChI key

CXBMCYHAMVGWJQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ. Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate. Download your certificate at: http://www.sigma-aldrich.com.

Tetramethrin, a type I pyrethroid, is used worldwide to protect crops and mammals from a variety of pests such as flies, fleas, mosquitoes, headlice, and mites. Tetramethrin is a synthetic pyrethroid insecticide, that binds to voltage-gated sodium channels of insect nerves and prevents its transition from an activated (ion-conducting) to an inactivated (non-conducting) state. This leads to a permanent depolarization of axonal membranes, causing paralysis and death of an insect.


Tetramethrin is not approved for use in the EU.
The maximum residue limits (MRLs) of pyrethroids compounds in fruits and fruit juices have been set at 0.02 to 0.5 mg/kg in China (GB2763-2019), and 0.02 to 0.3 mg/kg in European Union (EU) (Part A of Annex to Reg. 396/2005), respectively.

Application

It is intended to be used as a certified reference material (CRM) for calibration in chromatography and other analytical techniques. Tetramethrin in CRM may also find its use as described below:
  • Study of Estrogenic activity of tetramethrin in uterine CaBP-9k gene expression and a uterotrophic assay, and androgenic activity in Hershberger assay
  • To synthesize bis-dihydroxyboron fluorescein complexes for developing a ratiometric fluorescence probe used in the direct detection of pyrethroid residues in fruit juices
  • Separation of the four isomers of tetramethrin with ecotoxicity test and quantitative analysis in agrochemical formulations using chiral electrokinetic chromatography methodology
  • Residual detection of acetanilide, pentylenetetrazole, phenacetin, and tetramethrin in porcine muscle, pasteurized milk, and eggs by liquid chromatography-electrospray ionization tandem mass spectrometry (LC/ESI-MS/MS) method
  • To test glucocorticoid-like activity of propylparaben, butylparaben, diethylhexyl phthalate, and tetramethrin mixtures in different combinations at real human exposure levels

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - STOT SE 2 Inhalation

Target Organs

Nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dan Zhang et al.
Environmental monitoring and assessment, 188(6), 348-348 (2016-05-15)
A liquid chromatography-electrospray ionization tandem mass spectrometry (LC/ESI-MS/MS) method was developed for monitoring and detection of four different drugs, namely acetanilide, pentylenetetrazole, phenacetin, and tetramethrin in porcine muscle, pasteurized milk, and table egg samples. For acetanilide and pentylenetetrazole, the samples
M Greño et al.
The Science of the total environment, 800, 149496-149496 (2021-08-14)
The separation of the four isomers of tetramethrin was performed for the first time using a cyclodextrin-micellar electrokinetic chromatography methodology. Using sodium deoxycholate and 2-hydroxypropyl-β-CD as chiral selectors, tetramethrin isomers were separated with resolution values of 1.7 and 1.1 for
Dan Zhang et al.
Environmental monitoring and assessment, 188(6), 348-348 (2016-05-15)
A liquid chromatography-electrospray ionization tandem mass spectrometry (LC/ESI-MS/MS) method was developed for monitoring and detection of four different drugs, namely acetanilide, pentylenetetrazole, phenacetin, and tetramethrin in porcine muscle, pasteurized milk, and table egg samples. For acetanilide and pentylenetetrazole, the samples
Soon Sun Kim et al.
Journal of toxicology and environmental health. Part A, 68(23-24), 2277-2289 (2005-12-06)
Tetramethrin, a synthetic pyrethroid insecticide, is used globally for agriculture, and thus potential environmental exposure to tetramethrin is a concern. Environmental chemicals that are hormonally active (particularly estrogen or androgen) may adversely affect the reproductive and endocrine systems. However, little
Linda M Field et al.
European biophysics journal : EBJ, 46(7), 675-679 (2017-01-11)
The pyrethroid insecticides are a very successful group of compounds that have been used extensively for the control of arthropod pests of agricultural crops and vectors of animal and human disease. Unfortunately, this has led to the development of resistance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service