Skip to Content
Merck
All Photos(1)

Key Documents

59544-U

Supelco

SUPELCOSIL LC-ABZ (5 µm) HPLC Columns

L × I.D. 2 cm × 4 mm Supelguard Guard Cartridge pkg of 2 ea, Guard Cartridge holder required for use

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

Product Name

SUPELCOSIL LC-ABZ Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm

material

stainless steel column

Agency

suitable for USP L60

product line

SUPELCOSIL

feature

endcapped

packaging

pkg of 1 kit

technique(s)

HPLC: suitable

L × I.D.

2 cm × 4 mm

surface area

170 m2/g

matrix

fully porous particle

matrix active group

amide, alkyl phase

particle size

5 μm

pore size

120 Å

operating pH

2-7.5

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Footnote

Kit includes one cartridge, a stand-alone holder, a piece of tubing, and 2 nuts and ferrules.

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J G Ufkes et al.
Pharmacy world & science : PWS, 20(2), 83-87 (1998-05-19)
To study the pharmacokinetics of dextromoramide in long-term opiate addicts on methadone maintenance therapy (MMT) a reverse-phase HPLC technique was developed to monitor dextromoramide and methadone concentrations in plasma simultaneously. After liquid-liquid extraction from plasma, dextromoramide and methadone were determined
Neil C Dias et al.
The Analyst, 128(5), 427-433 (2003-06-07)
The solvation parameter model is used to identify suitable chromatographic models for estimating the octanol-water partition coefficient for neutral compounds of varied structure by reversed-phase liquid chromatography. The stationary phase Supelcosil LC-ABZ with methanol-water mobile phases affords a series of
Murali Pendela et al.
Journal of separation science, 32(20), 3418-3424 (2009-09-19)
Streptomycin (SM) is composed of streptidine, streptose and N-methyl glucosamine sugar moieties. For the determination of SM and its related substances, an ion-pair LC-UV detection method using a Supelcosil LC-ABZ column was developed previously. While analyzing commercial samples, several unknown
D Korakas et al.
Journal of pharmaceutical and biomedical analysis, 12(9), 1141-1145 (1994-09-01)
A series of biologically active peptides and their conjugates with lipidic amino acids were investigated by systematic change of the mobile phase composition using traditional octadecylsilica stationary phase and the newly developed Supelcosil LC-ABZ column. The mobile phases contained various
J A López et al.
Journal of chromatography. A, 870(1-2), 105-114 (2000-03-18)
A high-performance liquid chromatographic method with electrochemical detection has been developed for the determination of six 1,4-dihydropyridines: nifedipine, nimodipine, nisoldipine, nicardipine, felodipine and lacidipine. The chromatographic separation was performed using a Supelcosil LC-ABZ+Plus C18 column. A mobile phase of methanol-water

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service