Skip to Content
Merck
All Photos(1)

Key Documents

567504-U

Supelco

Discovery® HS F5 (3 µm) HPLC Columns

L × I.D. 5 cm × 4.6 mm, HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

product name

Discovery® HS F5 HPLC Column, 3 μm particle size, L × I.D. 5 cm × 4.6 mm

material

stainless steel column

Quality Level

Agency

suitable for USP L43

product line

Discovery®

feature

endcapped

manufacturer/tradename

Discovery®

packaging

1 ea of

extent of labeling

12% Carbon loading

parameter

≤70 °C temp. range
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable
LC/MS: suitable

L × I.D.

5 cm × 4.6 mm

surface area

300 m2/g

surface coverage

4 μmol/m2

impurities

<10 ppm metals

matrix

silica gel, high purity, spherical particle platform
fully porous particle

matrix active group

PFP (pentafluorophenyl) phase

particle size

3 μm

pore size

120 Å

operating pH range

2-8

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Guidelines for transferring a C18 method to Discovery® HS F5:
Generally, bases are retained longer on the HS F5 than on a C18. Increasing the organic content of a C18 separation 5 to 10 percent will generally provide similar retention on an HS F5. Results with other compounds are highly variable. However, it is generally true that solutes with log Po/w values less than 2.5 will be retained longer on HS F5 compared to a C18. The degree of difference is highly solute dependent.
The Discovery® HS F5 bonded phase provides reversed-phase separations that are distinctly different from C18 columns. However, compounds will generally elute within the same retention time window, making most C18 methods easily transferable.

Features and Benefits

  • Unique selectivity
  • Similar retention to C18 (sometimes requires stronger mobile phase)
  • Excellent peak shape
  • Stable, low-bleed LC-MS separations
  • Scalable separations from 3 to 10μm particle sizes

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Federica Pellati et al.
Journal of pharmaceutical and biomedical analysis, 48(2), 254-263 (2007-12-14)
In this study a pentafluorophenylpropyl (PFPP) stationary phase was applied to the fast and reliable qualitative and quantitative analysis of ephedrine alkaloids in Ephedra plant material and derivatives. A Discovery HS F5 column (150mmx4.6mm i.d., 5microm) was used, with an
Ilia Brondz et al.
Journal of pharmaceutical and biomedical analysis, 43(3), 937-944 (2006-11-03)
The drug primaquine diphosphate is used for causative treatment of malaria. Using HPLC-MS and GC-MS, this research group was previously able to show that the main contaminant of primaquine is the positional isomer quinocide [I. Brondz, D. Mantzilas, U. Klein
Koji Baba et al.
Journal of chromatography. A, 1354, 109-116 (2014-06-21)
Arsenic species, including arsenous acid, arsenic acid, methylarsonic acid, and dimethylarsinic acid, were determined using HPLC-ICPMS. The species were separated with a Discovery HS F5 column and a simple, volatile, and isocratic mobile phase of 0.1% (v/v) formic acid and
Federica Pellati et al.
Journal of chromatography. A, 1165(1-2), 58-66 (2007-08-07)
In this study, the chromatographic performance of a pentafluorophenylpropyl (PFPP) stationary phase was evaluated for the rapid separation of phenethylamine alkaloids (i.e. (+/-)-octopamine, (+/-)-synephrine, tyramine, N-methyltyramine and hordenine) in Citrus aurantium plant material (fruits and peel), various Citrus species, extracts
Jennie Wang et al.
Journal of pharmaceutical and biomedical analysis, 124, 267-273 (2016-03-13)
An HPLC method for the assay of a DNA topoisomerase inhibitor, LMP776 (NSC 725776), has been developed and validated. The stress testing of LMP776 was carried out in accordance with International Conference on Harmonization (ICH) guidelines Q1A (R2) under acidic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service