55511-U
Supel™ QuE QuEChERS tube
Z-Sep+ with MgSO4 Tube, pk of 50, centrifuge tube volume 15 mL , Shaker Compatible
Synonym(s):
QuEChERS
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
product name
Supel™ QuE, Z-Sep+ with MgSO4 Tube, pk of 50, centrifuge tube volume 15 mL , Shaker Compatible
composition
Magnesium Sulfate, 900 mg
Z-Sep+, 300 mg
packaging
pk of 50
technique(s)
QuEChERS: suitable
centrifuge tube volume
15 mL , Shaker Compatible
application(s)
food and beverages
Looking for similar products? Visit Product Comparison Guide
General description
Dispersive SPE (dSPE), often referred to as the "QuEChERS" method (Quick, Easy, Cheap, Effective, Rugged, and Safe), is modern sample prep technique that is becoming increasingly popular in the area of multi-residue pesticide analysis in food and agricultural products.
Using the QuEChERS method, food/agricultural samples are first extracted with an aqueous miscible solvent (e.g., acetonitrile) in the presence of high amounts of salts (e.g., sodium chloride and magnesium sulfate) and/or buffering agents (e.g. citrate) to induce liquid phase separation and stabilize acid and base labile pesticides, respectively. Upon shaking and centrifugation, an aliquot of the organic phase is subjected to further cleanup using SPE. Unlike traditional methods using SPE tubes, in dispersive SPE, cleanup is facilitated by mixing bulk amounts of SPE (e.g., Supelclean PSA, ENVI-Carb, and/or Discovery DSC-18) with the extract. After sample cleanup, the mixture is centrifuged and the resulting supernatant can either be analyzed directly or can be subjected to minor further treatment before analysis.
Supelco carries a line of vials and centrifuge tubes containing pre-determined amounts of salts and SPE sorbents to support the most common method configurations used today.
Using the QuEChERS method, food/agricultural samples are first extracted with an aqueous miscible solvent (e.g., acetonitrile) in the presence of high amounts of salts (e.g., sodium chloride and magnesium sulfate) and/or buffering agents (e.g. citrate) to induce liquid phase separation and stabilize acid and base labile pesticides, respectively. Upon shaking and centrifugation, an aliquot of the organic phase is subjected to further cleanup using SPE. Unlike traditional methods using SPE tubes, in dispersive SPE, cleanup is facilitated by mixing bulk amounts of SPE (e.g., Supelclean PSA, ENVI-Carb, and/or Discovery DSC-18) with the extract. After sample cleanup, the mixture is centrifuged and the resulting supernatant can either be analyzed directly or can be subjected to minor further treatment before analysis.
Supelco carries a line of vials and centrifuge tubes containing pre-determined amounts of salts and SPE sorbents to support the most common method configurations used today.
Compatibility
Compatible with most QuEChERS shakers.
Other Notes
Supel™ QuE Z-Sep products enhance sample cleanup for complex matrices by effectively removing more fat and color from sample extracts than traditional phases for QuEChERS methods. By eliminating problematic matrix interferences, Z-Sep products provide more robust LC-MS and GC-MS methods. This proprietary technology can replace C18 and PSA in your current methods without additional method development.
Z-Sep+ is recommended for cleanup of samples containing greater than 15% fat.
Z-Sep/C18 is recommended for cleanup of samples containing less than 15% fat.
Z-Sep is recommended for cleanup of samples prior to analysis of hydrophobic analytes.
Z-Sep+ is recommended for cleanup of samples containing greater than 15% fat.
Z-Sep/C18 is recommended for cleanup of samples containing less than 15% fat.
Z-Sep is recommended for cleanup of samples prior to analysis of hydrophobic analytes.
Legal Information
Certain uses of this product may be covered by the claims of one or more pending patent applications. For licensing inquiries please contact techserv@sial.com
AOAC is a registered trademark of AOAC International
Supel is a trademark of Sigma-Aldrich Co. LLC
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Journal of chromatography. A, 1304, 109-120 (2013-07-23)
Several extraction methods were evaluated in terms of recoveries and extraction precision for 113 pesticides in avocado: QuEChERS with various d-SPE clean-ups (Z-Sep, Z-Sep+, PSA+C18 and silica), miniLuke and ethyl acetate. Extracts were analysed using liquid chromatography coupled with triple
Journal of chromatography. A, 1339, 34-41 (2014-03-25)
A Quick, Easy, Cheap, Effective, Rugged and Safe (QuEChERS) extraction method followed by liquid chromatography-(Orbitrap) high resolution mass spectrometry was developed for the simultaneous determination of five benzotriazole, four benzothiazole and five benzenesulfonamide derivates in sewage sludge. While the method
Journal of AOAC International, 100(6), 1671-1680 (2017-07-15)
In this study, a simple, cost-effective, and sensitive HPLC diode-array detection method was developed for the simultaneous determination of six different 5-nitroimidazoles [metronidazole, 2-hydroxymethyl-1-methyl-5-nitro-1H-imidazole, dimetridazole (DMZ), ronidazole, ornidazole, and ipronidazole] in bovine milk samples. A QuEChERS-based sample preparation procedure was
Journal of chromatography. A, 1528, 53-60 (2017-11-07)
Herein, an accurate and reliable isotope-labelled internal standard method was developed and validated for simultaneous determination of three polar pesticide adjuvants, namely 2-pyrrolidone, N-methyl-2-pyrrolidone, and N-ethyl-2-pyrrolidone in plant-derived agro-products. Matrices, including apple, cabbage, tomato, cucumber, rice, and wheat were extracted
Talanta, 118, 68-83 (2013-11-28)
Two sorbents containing ZrO₂ (Z-Sep and Z-Sep+) were tested as a d-SPE clean-up in combination with the QuEChERS and ethyl acetate multiresidue method in the pesticide residues extraction in avocado. All extracts were analysed using gas chromatography coupled with a
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service