Skip to Content
Merck
All Photos(1)

Documents

48597

Supelco

cis-1,2-Dichloroethene

analytical standard

Synonym(s):

cis-1,2-Dichloroethylene, cis-Acetylene dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH=CHCl
CAS Number:
Molecular Weight:
96.94
Beilstein:
1071208
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Quality Level

CofA

current certificate can be downloaded

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.449 (lit.)

bp

60 °C (lit.)

mp

−80 °C (lit.)

density

1.284 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-8°C

SMILES string

Cl\C=C/Cl

InChI

1S/C2H2Cl2/c3-1-2-4/h1-2H/b2-1-

InChI key

KFUSEUYYWQURPO-UPHRSURJSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

recommended

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biao Jin et al.
Environmental science & technology, 48(11), 6141-6150 (2014-05-09)
Laboratory experiments were performed to investigate and quantify the extent of diffusive isotope fractionation of organic contaminants in aqueous solution. We selected petroleum hydrocarbons (toluene and ethylbenzene, in 1:2 mixtures of labeled (perdeuterated) and nonlabeled isotopologues) and chlorinated solvents (trichloroethene
Jaejin Lee et al.
Biodegradation, 22(4), 687-698 (2010-11-06)
Tetrachloroethene (PCE) and trichloroethene (TCE) are common groundwater contaminants that also impact tidal flats, especially near urban and industrial areas. However, very little is known about dechlorinating microbial communities in tidal flats. Titanium pyrosequencing, 16S rRNA gene clone libraries, and
Wang Suhua et al.
Archives of toxicology, 84(6), 461-469 (2010-02-04)
The present study was designed to examine the effects of the inhibition or induction of CYP2E1 activity on acute acrylonitrile (AN) toxicity in rats. Increased or decreased hepatic CYP2E1 activity was achieved by pretreatment with acetone or trans-1,2-dichloroethylene (DCE), respectively.
Daniel Hunkeler et al.
Environmental science & technology, 45(19), 8247-8253 (2011-08-30)
Compound-specific isotope analysis (CSIA) can potentially be used to relate vapor phase contamination by volatile organic compounds (VOCs) to their subsurface sources. This field and modeling study investigated how isotope ratios evolve during migration of gaseous chlorinated ethenes across a
Daniel Hunkeler et al.
Journal of contaminant hydrology, 119(1-4), 69-79 (2010-10-30)
The fate of chlorinated ethenes in a large contaminant plume originating from a tetrachloroethene (PCE) source in a sandy aquifer in Denmark was investigated using novel methods including compound-specific carbon and chlorine isotope analysis and quantitative real-time polymerase chain reaction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service