Skip to Content
Merck
All Photos(1)

Key Documents

48499

Supelco

Indeno[1,2,3-cd]pyrene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H12
CAS Number:
Molecular Weight:
276.33
Beilstein:
1879312
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 10 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

c1ccc-2c(c1)-c3ccc4ccc5cccc6cc-2c3c4c56

InChI

1S/C22H12/c1-2-7-17-16(6-1)18-11-10-14-9-8-13-4-3-5-15-12-19(17)22(18)21(14)20(13)15/h1-12H

InChI key

SXQBHARYMNFBPS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Indeno[1,2,3-cd]pyrene is a polycyclic aromatic hydrocarbon, which is a major environmental pollutant. It can be formed via incomplete combustion of organic matter.

Application

Indeno[1,2,3-cd]pyrene may be used as an analytical standard for the determination of the analyte in sewage sludge, solvent solutions and food extracts, water, and oil samples by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Ion Mobility: TWCCSN2 value of 154.8 Å2 [M+H]+

The collision cross section (CCS) measurement was provided by Waters Corporation, using the SYNAPT XS mass spectrometer.
For a description and overview of how ion mobility enables the measurement of the CCS of an ion visit ims.waters.com.
Further information on the SYNAPT XS mass spectrometer can be found on the IMS microsite and product webpage.

TWCCS measurements are expected to be within 2% of this reference value.

P/N 48499 is part of the Waters Extractables & Leachables UNIFI scientific library which can be downloaded from Waters Marketplace.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

PAH source fingerprints for coke ovens, diesel and, gasoline engines, highway tunnels, and wood combustion emissions
Khalili.RM, et al.
Atmospheric Environment, 29, 533-542 (1995)
M Villar et al.
Analytica chimica acta, 599(1), 92-97 (2007-09-04)
A new approach has been developed for the monitoring of linear alkylbenzene sulfonates (LAS) (C10-C13) in sewage sludge. It is based on microwave-assisted extraction (MAE) prior to high performance liquid chromatography coupled with diode array (HPLC-DAD) and fluorescence (HPLC-FL) detectors.
Polycyclic aromatic hydrocarbons in the diet
Phillips.HD
Mutation Research. Genetic Toxicology and Environmental Mutagenesis, 443, 139-147 (1999)
Identification of mutagenic metabolites of indeno [1, 2, 3-cd] pyrene formed in vitro with rat liver enzymes
Rice.EJ, et al.
Cancer Research, 45, 5421-5425 (1985)
Francisco Galán-Cano et al.
Journal of chromatography. A, 1300, 134-140 (2013-04-23)
In this paper, ionic liquid coated magnetic nanoparticles (IL-MNPs) have been prepared by covalent immobilization. The as-synthesized MNPs have been successfully used as sorbent for the extraction of polycyclic aromatic hydrocarbons (PAHs) from water samples, the analytes being finally determined

Articles

This application note describes the fast and efficient separation of the EU’s list of 15 + 1 polynuclear aromatic hydrocarbons (PAHs) using Ascentis® Express PAH HPLC column.

This application note describes the fast and efficient separation of the EU’s list of 15 + 1 polynuclear aromatic hydrocarbons (PAHs) using Ascentis® Express PAH HPLC column.

This application note describes the fast and efficient separation of the EU’s list of 15 + 1 polynuclear aromatic hydrocarbons (PAHs) using Ascentis® Express PAH HPLC column.

This application note describes the fast and efficient separation of the EU’s list of 15 + 1 polynuclear aromatic hydrocarbons (PAHs) using Ascentis® Express PAH HPLC column.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service