Skip to Content
Merck
All Photos(1)

Key Documents

47778

Supelco

EPA 8260A Surrogate Standards Mix

certified reference material, 2000 μg/mL each component in methanol

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

Agency

EPA 8260,8260A

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type surrogate

packaging

ampule of 1 mL

concentration

2000 μg/mL each component in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    4-Bromofluorobenzene
    Dibromofluoromethane
    1,2-Dichloroethane-d4
    Toluene-d8

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Flam. Liq. 2 - Ozone 1 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammad Bakhtiar Hesam Shariati et al.
Journal of cellular physiology, 234(11), 20098-20110 (2019-04-11)
Implantation of embryos needs endometrial receptivity. Mineralocorticoids is one of the causes influencing the implantation window. This study targeted to evaluation fludrocortisone different properties on endometrial receptivity. The objective of this study was to assess whether treatment with fludrocortisone could
Jasmin Felux et al.
Frontiers in immunology, 12, 613597-613597 (2021-03-23)
Increased concentrations of circulating chromatin, especially oligo-nucleosomes, are observed in sepsis, cancer and some inflammatory autoimmune diseases like systemic lupus erythematosus (SLE). In SLE, circulating nucleosomes mainly result from increased apoptosis and decreased clearance of apoptotic cells. Once released, nucleosomes
Won Hyeok Lee et al.
Cells, 9(8) (2020-08-14)
Tumor necrosis factor (TNF)-related apoptosis-inducing ligand (TRAIL) has attracted attention as a potential candidate for cancer therapy. However, many primary cancers are resistant to TRAIL, even when combined with standard chemotherapy. The mechanism of TRAIL resistance in cancer cells has
Sonal Srikanth et al.
Nature immunology, 20(2), 152-162 (2019-01-16)
Stimulator of interferon genes (STING) is an endoplasmic reticulum (ER) signaling adaptor that is essential for the type I interferon response to DNA pathogens. Aberrant activation of STING is linked to the pathology of autoimmune and autoinflammatory diseases. The rate-limiting
Zhouwulin Shao et al.
Viruses, 11(12) (2019-12-01)
Autophagy is an essential catabolic process that degrades cytoplasmic components within the lysosome, therefore ensuring cell survival and homeostasis. A growing number of viruses, including members of the Herpesviridae family, have been shown to manipulate autophagy to facilitate their persistence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service