24407
SGE Syringes for PerkinElmer® Autosystem
volume 0.5 μL, 0.5BR-PE-0.63, removable needle, needle size 23 ga, needle L 70 mm
All Photos(1)
About This Item
UNSPSC Code:
41122001
Recommended Products
material
cone tip stainless steel needle (point style 1)
needle size
23 ga
description
color (light orange)
liquid syringe
feature
removable needle
packaging
pkg of 1 ea
manufacturer/tradename
SGE 000478
needle L
70 mm
volume
0.5 μL
Legal Information
PerkinElmer is a registered trademark of PerkinElmer Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)