Key Documents
22781
Supelco
Fritted Purge Sampler
size 5 mL, Tekmar® 14-2337-024
About This Item
UNSPSC Code:
41115700
material
glass
Quality Level
Agency
suitable for EPA 601
manufacturer/tradename
Tekmar® 14-2337-024
size
5 mL
application(s)
industrial hygiene
compatibility
for use with Tekmar® 2000/3000
for use with standard aqueous analysis
Features and Benefits
Note: All 1/4 in. O.D. glass ends are ground glass.
Legal Information
Tekmar is a registered trademark of Tekmar Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Visit the Document Library
Related Content
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.