Skip to Content
Merck
All Photos(1)

Key Documents

12168-U

Supelco

Tenax® Porous Polymer Adsorbent

matrix Tenax TA (refined), 60-80 mesh, bottle of 100g

Synonym(s):

stopcock (3-way), luer lock, polycarbonate/acetal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100
NACRES:
SB.54

Product Name

Tenax® Porous Polymer Adsorbent, matrix Tenax TA (refined), 60-80 mesh, bottle of 100 g

Agency

suitable for EPA 601

Quality Level

form

solid

packaging

bottle of 100 g

parameter

350 °C temp. range

surface area

~35 m2/g

matrix

Tenax TA (refined)

particle size

60-80 mesh

bulk density

~0.25 g/mL (free fall density)

Looking for similar products? Visit Product Comparison Guide

General description

Tenax porous polymers, based on 2,6-diphenyl-p-phenylene oxide, is widely used as an adsorbent in both air collection and purge trap applications. Its unique structure provides alternate and desirable adsorption/desorption characteristics compared to other porous polymers.

For more information about any of our adsorbents, please visit sigma-aldrich.com/adsorbents

Application


  • Optimization of VOC Sampling: Research on Tenax® porous polymer adsorbents has demonstrated their efficacy in sampling very volatile organic compounds (VOCs) in both dry and humid air, crucial for environmental testing. This study outlines the method for determining recovery rates using thermal desorption gas chromatography-mass spectrometry, highlighting Tenax®′s role in achieving high-precision analyses at sub-ppb levels (Richter et al., 2020).

  • Enhanced Gas Chromatography Performance: A comparative analysis of Tenax® and another common adsorbent material for thermal desorption gas chromatography - mass spectrometry of biogenic volatile organic compounds underscores Tenax®′s superior performance. The study emphasizes its high surface area and chemical stability, making it ideal for complex sample matrices in pharmaceutical and environmental applications (Marcillo et al., 2017).

Footnote

This material is refined (screened) then repackaged.

Legal Information

Tenax is a registered trademark of Buchem B.V.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Fire Investigation
Daeid NN.
Science, 2004, 142-143 null
Static Headspace-Gas Chromatography: Theory and Practice .
Kolb B and Ettre LS.
Science, 6-7 (2006)
Handbook of Water Analysis.
Nollet LML and Gelder LSP.
Technology & Engineering, 2007, 625-626 null
Evaluation of Anasorb CMS and comparison with Tenax TA for the sampling of volatile organic compounds in indoor and outdoor air by breakthrough measurements.
Maria P.
Analyst, 121 (3), 303-307 (1996)

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service