Skip to Content
Merck
All Photos(1)

Key Documents

Z740213

Supelco

TLC Glass Plates

L × W 2.5 cm × 7.5 cm, silica gel 60 matrix, binder, Organic Polymer, fluorescent indicator

Synonym(s):

TLC Silica gel 60 F254, TLC Silica gel 60 F254, 100 Glass plates 2.5 x 7.5 cm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
NACRES:
NB.21
Pricing and availability is not currently available.

material

glass support
silica gel 60 matrix

Quality Level

feature

binder Organic Polymer
fluorescent indicator

packaging

pkg of 100 ea

manufacturer/tradename

EMD Millipore® 1.15327.0001

technique(s)

thin layer chromatography (TLC): suitable

L × W

2.5 cm × 7.5 cm

layer thickness

250 μm

particle size

10-12 μm

pore size

60 Å medium pore diameter

Looking for similar products? Visit Product Comparison Guide

General description

Our classical silica TLC plates are based on a combination of our proven silica gel 60 and the addition of a unique polymeric binder resulting in a very adherent and hard surface that will not crack or blister and even allow writing with a pensil on the surface without risk to damage the layer.

Footnote

Product packaged in a plastic box

Legal Information

EMD Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service