Z734640
Nunc® Lab-Tek® II chambered coverglass
2 wells, polystyrene chambers, 1.5 borosilicate coverglass, 4.0 cm2/well, 96/cs
Synonym(s):
chamber slides, microscope slides, microscopy slides
About This Item
material
borosilicate glass
colorless
polystyrene chamber
sterility
sterile
quality
CE marked
packaging
case of 96
manufacturer/tradename
Nunc 155379
W × L
24 mm × 55 mm
well area
4 cm2
wells
2
working volume
1.0-2.0 mL
suitability
suitable for (high power inverted microscopic viewing; confocal image analysis)
Looking for similar products? Visit Product Comparison Guide
General description
- Excellent for confocal image analysis
- Optimal for high-power inverted microscopic viewing
- Chambered coverglass
- The medium chamber is not removable
Application
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service