Skip to Content
Merck
All Photos(1)

Key Documents

Z709107

ART® non-filtered tip

HLT 1000 SoftFit, maximum volume 1000 μL, racked, non-sterile

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.01

sterility

non-sterile

feature

DNase free
RNase free
racked

packaging

pack of 800 ea

manufacturer/tradename

Molecular BioProducts 3582

technique(s)

PCR: suitable

maximum volume

1000 μL

Looking for similar products? Visit Product Comparison Guide

General description

Molecular BioProducts Pure lines of pipette tips are pre-sterilized and packaged in a controlled environment to eliminate contamination. These tips are tested by an independent lab to ensure that they meet industry standards. It is ideal for a wide variety of applications.
SoftFit design makes loading and ejecting easy and provides a universal fit.

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.
SoftFit-L is a trademark of Thermo Fisher Scientific or its subsidiaries

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service