Skip to Content
Merck
All Photos(1)

Key Documents

Z4527

Sigma-Aldrich

Zopolrestat

≥98% (HPLC)

Synonym(s):

3,4-Dihydro-4-oxo-3-[[5-(trifluoromethyl)-2-benzothiazolyl]methyl]-1-phthalazineacetic acid, Alond, CP-73850, Xedia

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H12F3N3O3S
CAS Number:
Molecular Weight:
419.38
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

off-white to light brown

solubility

DMSO: ≥20 mg/mL

storage temp.

2-8°C

SMILES string

OC(=O)CC1=NN(Cc2nc3cc(ccc3s2)C(F)(F)F)C(=O)c4ccccc14

InChI

1S/C19H12F3N3O3S/c20-19(21,22)10-5-6-15-14(7-10)23-16(29-15)9-25-18(28)12-4-2-1-3-11(12)13(24-25)8-17(26)27/h1-7H,8-9H2,(H,26,27)

InChI key

BCSVCWVQNOXFGL-UHFFFAOYSA-N

Biochem/physiol Actions

Zopolrestat is an inhibitor of Aldose Reductase. AR family members AKR1B1 and AKR1B10 have additionally been shown to play roles in inflammation and cancer.

Features and Benefits

This compound is featured on the Dopamine and Norepinephrine Metabolism page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cornelia Koch et al.
Biochimica et biophysica acta, 1810(9), 879-887 (2011-06-21)
Flexibility is a common feature of proteins. For human aldose reductase, a variety of conformers have been observed in crystalline complexes with different inhibitors. A study of crystal structures and isothermal titration calorimetry was performed on wild type and mutated
Liping Zou et al.
International journal of molecular medicine, 30(2), 409-416 (2012-05-23)
Aldose reductase (AR), the first and the rate-limiting enzyme of the polyol pathway, has been implicated in platelet-derived growth factor (PDGF)-induced proliferation of rat mesangial cells (MsCs). It is well known that AR plays an important role in various chronic
Holger Steuber et al.
Journal of molecular biology, 363(1), 174-187 (2006-09-06)
In structure-based drug design, accurate crystal structure determination of protein-ligand complexes is of utmost importance in order to elucidate the binding characteristics of a putative lead to a given target. It is the starting point for further design hypotheses to
Depeng Jiang et al.
International immunopharmacology, 12(4), 588-593 (2012-03-06)
This study investigated whether aldose reductase (AR) inhibition affects interleukin (IL)-13-induced mucus production in the human bronchial epithelial cell line-16 (HBE16) cells. The HBE16 cells were cultured with AR inhibitors (zopolrestat) or were transfected with an AR small interfering (si)RNA.
Brian F Johnson et al.
Diabetes care, 27(2), 448-454 (2004-01-30)
The goal of this study was to determine whether treatment with an aldose reductase inhibitor (ARI) has beneficial effects on asymptomatic cardiac abnormalities in diabetic patients with neuropathy. Diabetic subjects with neuropathy (n = 81) with either a low diastolic

Articles

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service