Z325139
BRAND® BLAUBRAND® ETERNA volumetric pipette, calibrated To Deliver
capacity 5 mL
About This Item
Recommended Products
material
AR-Glas®
Agency
according to DIN 12 691
description
amber mark: yes
feature
class AS with batch number
packaging
pack of 6 ea
manufacturer/tradename
BRAND 30507
volume measuring range
5 mL, accuracy: 0.015 mL
L
400 mm
capacity
5 mL
Looking for similar products? Visit Product Comparison Guide
General description
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
This page contains detailed information on working with and handling pipettes and pipetting aids.
This page contains detailed information on working with and handling pipettes and pipetting aids.
This page contains detailed information on working with and handling pipettes and pipetting aids.
This page contains detailed information on working with and handling pipettes and pipetting aids.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service