Skip to Content
Merck
All Photos(1)

Key Documents

Z2001

Sigma-Aldrich

Zonisamide sodium salt

≥98% (HPLC), powder

Synonym(s):

1,2-Benzisoxazole-3-methanesulfonamide sodium-potassium salt, Aleviatin sodium-potassium salt, Exceglan sodium-potassium salt, Excegram sodium-potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7N2NaO3S
CAS Number:
Molecular Weight:
234.21
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: >5 mg/mL

originator

Eisai

SMILES string

[Na]NS(=O)(=O)Cc1noc2ccccc12

InChI

1S/C8H7N2O3S.Na/c9-14(11,12)5-7-6-3-1-2-4-8(6)13-10-7;/h1-4H,5H2,(H-,9,11,12);/q-1;+1

InChI key

ZVBIRPKGWOVBLG-UHFFFAOYSA-N

Biochem/physiol Actions

Zonisamide sodium salt is an anti-epileptic and also scavenges nitric oxide (NO).
Zonisamide sodium salt is an anti-epileptic. It is effective in various animal epilepsy models and humans with both partial and generalized epileptic seizures. Zonisamide sodium salt also scavenges nitric oxide (NO).

Features and Benefits

This compound was developed by Eisai. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zonisamide for the treatment of self-inflicted dermatoses related to impulse control disorders.
Maria José Tribó et al.
Archives of dermatology, 148(6), 762-764 (2012-06-20)
Vedat Ali Yürekli et al.
Cellular and molecular neurobiology, 33(2), 205-212 (2012-12-12)
Parkinson's disease is an incurable progressive neurological condition caused by a degeneration of dopamine-producing cells characterized by motor and non-motor symptoms. The major mechanisms of the antiepileptic actions of ZNS are inhibition of voltage-gated Na(+) channel, T-type voltage-sensitive Ca(2+) channel
Kouji Fukuyama et al.
Pharmaceuticals (Basel, Switzerland), 13(5) (2020-05-24)
To understand the pathomechanism and pathophysiology of autosomal dominant sleep-related hypermotor epilepsy (ADSHE), we studied functional abnormalities of glutamatergic transmission in thalamocortical pathway from reticular thalamic nucleus (RTN), mediodorsal thalamic nucleus (MDTN) to orbitofrontal cortex (OFC) associated with S286L-mutant α4β2-nicotinic
Dong Wook Kim et al.
Epilepsy & behavior : E&B, 23(4), 497-499 (2012-03-24)
Because iatrogenic obesity may hinder medication compliance, it would be a reasonable approach to consider antiepileptic drugs (AEDs) that promote weight loss in overweight patients. We performed an open-label, observational study to assess the effects of zonisamide on weight in
J Y Chung et al.
New Zealand veterinary journal, 60(6), 357-359 (2012-05-30)
To evaluate the efficacy of zonisamide as a monotherapy in dogs with idiopathic epileptic seizure. The experiment was conducted on 10 dogs with idiopathic epilepsy that were treated at the Seoul National University Hospital for Animals. A diagnosis was conducted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service