Skip to Content
Merck
All Photos(1)

Key Documents

X4626

Sigma-Aldrich

XTT sodium salt

powder, BioReagent, suitable for cell culture

Synonym(s):

2,3-Bis(2-methoxy-4-nitro-5-sulfophenyl)-2H-tetrazolium-5-carboxanilide inner salt, 2,3-Bis(2-methoxy-4-nitro-5-sulfophenyl)-2H-tetrazolium-5-carboxanilide inner salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H16N7NaO13S2
CAS Number:
Molecular Weight:
673.52
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.75

product line

BioReagent

Quality Level

Assay

≥90%

form

powder

storage condition

dry at room temperature

technique(s)

cell culture | mammalian: suitable
protein quantification: suitable

application(s)

detection

detection method

colorimetric

storage temp.

2-8°C

SMILES string

[Na+].COc1cc(c(cc1-n2nc(n[n+]2-c3cc(c(cc3OC)[N+]([O-])=O)S([O-])(=O)=O)C(=O)Nc4ccccc4)S([O-])(=O)=O)[N+]([O-])=O

InChI

1S/C22H17N7O13S2.Na/c1-41-17-8-15(28(31)32)19(43(35,36)37)10-13(17)26-24-21(22(30)23-12-6-4-3-5-7-12)25-27(26)14-11-20(44(38,39)40)16(29(33)34)9-18(14)42-2;/h3-11H,1-2H3,(H2-,23,30,35,36,37,38,39,40);/q;+1/p-1

InChI key

JACYMBNQPPWQML-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

XTT (sodium 3′-[1-[(phenylamino)-carbony]-3,4-tetrazolium]-bis(4-methoxy-6-nitro)benzene-sulfonic acid hydrate) is a tetrazolium salt, which forms water-soluble formazan on bioreduction.

Application

XTT has been used in conjunction with phenazine methosulfate (PMS) to screen human cancer cell lines. The tetrazolium dye has also been used to study fungal cell damage, in testing antimicrobial susceptibility of staphylococci, and in Candida biofilm research. Limitations to the XTT assay have been reported.
XTT sodium salt has been used to assess cell viability.

Biochem/physiol Actions

XTT (sodium 3′-[1-[(phenylamino)-carbony]-3,4-tetrazolium]-bis(4-methoxy-6-nitro)benzene-sulfonic acid hydrate) is acted upon by dehydrogenases present in metabolically active cells. This produces water soluble formazan, which is a highly colored product. Thus, it offers an advantage in colorimetric proliferation assays, as opposed to MTT, as the product is already soluble and no formazan crystals are formed.

Principle

In living cells, XTT is metabolically reduced to produce a colorimetric, water-soluble formazan product.

related product

Product No.
Description
Pricing

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-react. C

Supplementary Hazards

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antimicrobial activity, cytotoxicity and chemical analysis of lemongrass essential oil (Cymbopogon flexuosus) and pure citral
Adukwu EC, et al.
Applied Microbiology and Biotechnology, 100(22), 9619-9627 (2016)
Amos C Hung et al.
Oncology letters, 22(5), 774-774 (2021-10-01)
Esophageal cancer is one of the most common malignancies and leading cause of cancer-associated mortality worldwide. However, the molecular mechanisms underlying esophageal cancer progression and the development of clinical tools for effective diagnosis remain unclear. Resistin, which was originally identified
Jesús Gandara-Loe et al.
ACS applied materials & interfaces, 11(2), 1924-1931 (2018-12-19)
Metal-organic frameworks (MOFs) have been evaluated as potential nanocarriers for intraocular incorporation of brimonidine tartrate to treat chronic glaucoma. Experimental results show that UiO-67 and MIL-100 (Fe) exhibit the highest loading capacity with values up to 50-60 wt %, whereas
L M Jost et al.
Journal of immunological methods, 147(2), 153-165 (1992-03-04)
A tetrazolium compound, XTT, bioreducible to a water-soluble formazan was used to develop a simplified cellular cytotoxicity assay. Most (13/15 melanoma and 2/3 colon carcinoma cell lines tested metabolized XTT greater than 50 times more efficiently than the lymphoid effector
Wander José da Silva et al.
Brazilian dental journal, 19(4), 364-369 (2009-01-31)
2, 3-bis (2-methoxy-4-nitro-5-sulfophenyl)-5-[(phenylamino) carbonyl]-2H-tetrazolium hydroxide (XTT) reduction assay has been used to study Candida biofilm formation. However, considering that the XTT reduction assay is dependent on cell activity, its use for evaluating mature biofilms may lead to inaccuracies since biofilm

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service