Skip to Content
Merck
All Photos(7)

Key Documents

WH0009927M3

Sigma-Aldrich

Monoclonal Anti-MFN2 antibody produced in mouse

clone 4H8, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-CMT2A, Anti-CMT2A2, Anti-CPRP1, Anti-HSG, Anti-KIAA0214, Anti-MARF, Anti-mitofusin 2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA (i)
IHC (p)
WB
clone:
4H8, monoclonal
species reactivity:
human, mouse, rat
citations:
17
technique(s):
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4H8, monoclonal

form

buffered aqueous solution

species reactivity

human, mouse, rat

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MFN2(9927)

General description

This gene encodes a mitochondrial membrane protein that participates in mitochondrial fusion and contributes to the maintenance and operation of the mitochondrial network. This protein is involved in the regulation of vascular smooth muscle cell proliferation, and it may play a role in the pathophysiology of obesity. Mutations in this gene cause Charcot-Marie-Tooth disease type 2A2, and hereditary motor and sensory neuropathy VI, which are both disorders of the peripheral nervous system. Defects in this gene have also been associated with early-onset stroke. Two transcript variants encoding the same protein have been identified. (provided by RefSeq)

Immunogen

MFN2 (NP_055689, 661 a.a. ~ 757 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
FKRQFVEHASEKLQLVISYTGSNCSHQVQQELSGTFAHLCQQVDVTRENLEQEIAAMNKKIEVLDSLQSKAKLLRNKAGWLDSELNMFTHQYLQPSR

Biochem/physiol Actions

Mitofusin 2 (MFN2) plays an essential role in mitochondrial metabolism via maintenance and operation of the mitochondrial network architecture through fusion of mitochondria. Decreased expression of MFN2 leads to metabolic abnormalities, which further increases the risk of developing obesity and type 2 diabetes. Mutation of the gene results in Charcot–Marie–Tooth neuropathy type 2A (CMT2A). Mfn2 controls various cellular function including cell proliferation, oxidative metabolism, autophagy, and mitochondrial antiviral signaling protein. MFN2 integrates mitochondria and endoplasmic reticulum function, which is required for regulation of insulin signaling and glucose homeostasis in vivo. Repressed expression of MFN2 in placenta is associated with pathogenesis of preeclampsia (PE). MFN2 functions as a biomarker and therapeutic target for PE and cardiovascular diseases, such as hypertension.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhihao Wei et al.
International journal of biological sciences, 19(6), 1925-1940 (2023-04-18)
Background: Fatty acid oxidation (FAO) is a major alternate energy metabolism pathway in tumor cells subjected to metabolic stress caused by glucose deficiency during rapid progression. However, the mechanism of metabolic reprogramming between glycolysis and FAO in tumor cells is
Du Fang et al.
Scientific reports, 6, 31462-31462 (2016-08-19)
Mitochondria are essential dynamic organelles for energy production. Mitochondria dynamically change their shapes tightly coupled to fission and fusion. Imbalance of fission and fusion can cause deficits in mitochondrial respiration, morphology and motility. Mfn2 (mitofusin 2), a mitochondrial membrane protein
Abdennour Douida et al.
International journal of molecular sciences, 22(4) (2021-02-11)
The conserved Blm10/PA200 proteins are proteasome activators. Previously, we identified PA200-enriched regions in the genome of SH-SY5Y neuroblastoma cells by chromatin immunoprecipitation (ChIP) and ChIP-seq analysis. We also found that selective mitochondrial inhibitors induced PA200 redistribution in the genome. Collectively
Jun Yu et al.
BioMed research international, 2016, 6323086-6323086 (2016-03-05)
Background. Mitofusin 2 (Mfn2) is a novel mitochondrial protein that is implicated in cellular proliferation and metabolism; however, the role of Mfn2 in preeclampsia (PE) remains unknown. This study aimed to explore the relationship between Mfn2 and PE. Method. Preeclamptic
Mutations in the mitochondrial GTPase mitofusin 2 cause Charcot-Marie-Tooth neuropathy type 2A.
Stephan Züchner et al.
Nature genetics, 36(5), 449-451 (2004-04-06)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service