Skip to Content
Merck
All Photos(2)

Key Documents

WH0003848M1

Sigma-Aldrich

Monoclonal Anti-KRT1 antibody produced in mouse

clone 2A7, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-CK1, Anti-EHK1, Anti-K1, Anti-KRT1A, Anti-keratin 1 (epidermolytic hyperkeratosis)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2A7, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KRT1(3848)

General description

Keratin 1 (KRT1) is a cytoplasmic intermediate filament protein, that is expressed in the basal layer of the epidermis. This gene is located on human chromosome 12q13.
The protein encoded by this gene is a member of the keratin gene family. The type II cytokeratins consist of basic or neutral proteins which are arranged in pairs of heterotypic keratin chains coexpressed during differentiation of simple and stratified epithelial tissues. This type II cytokeratin is specifically expressed in the spinous and granular layers of the epidermis with family member KRT10 and mutations in these genes have been associated with bullous congenital ichthyosiform erythroderma. The type II cytokeratins are clustered in a region of chromosome 12q12-q13. (provided by RefSeq)

Immunogen

KRT1 (NP_006112, 387 a.a. ~ 496 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
HGDSVRNSKIEISELNRVIQRLRSEIDNVKKQISNLQQSISDAEQRGENALKDAKNKLNDLEDALQQAKEDLARLLRDYQELMNTKLALDLEIATYRTLLEGEESRMSGE

Biochem/physiol Actions

Keratin 1 (KRT1) helps to maintain skin integrity and participates in inflammatory response. It plays an important role in the differentiation and function of keratinocytes. Since KRT1 is one of the important cytoskeleton filament, it extends mechanical support and participates in additional activities in epithelial cells.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Allele-specific KRT1 expression is a complex trait
Tao H, et al.
PLoS Genetics, 2(6), e93-e93 (2006)
Polymorphism of keratin 1 associates with systemic lupus erythematosus and systemic sclerosis in a south Chinese population
Luo W, et al.
PLoS ONE, 12(10) (2017)
Keith A Choate et al.
The Journal of clinical investigation, 125(4), 1703-1707 (2015-03-17)
Widespread reversion of genetic disease is rare; however, such events are particularly evident in some skin disorders in which normal clones develop on a background of affected skin. We previously demonstrated that mutations in keratin 10 (KRT10) cause ichthyosis with
Linkage of the gene for the triple A syndrome to chromosome 12q13 near the type II keratin gene cluster
Weber A, et al.
Human Molecular Genetics, 5(12), 2061-2066 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service