Skip to Content
Merck
All Photos(6)

Key Documents

WH0002119M2

Sigma-Aldrich

Monoclonal Anti-ETV5 antibody produced in mouse

clone 7C10, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-ERM, Anti-ets variant gene 5 (ets-related molecule)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7C10, monoclonal

form

buffered aqueous solution

species reactivity

human, mouse, rat

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ETV5(2119)

General description

E-twenty-six (Ets) variant gene 5 (ETV5) protein belongs to the polyoma enhancer activator 3 (PEA3) subfamily of ETS transcription factors. The ETV5 gene is located on the human chromosome at 3q27.2.

Immunogen

ETV5 (NP_004445, 181 a.a. ~ 290 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
LPEPGPQQQTFAVPRPPHQPLQMPKMMPENQYPSEQRFQRQLSEPCHPFPPQPGVPGDNRPSYHRQMSEPIVPAAPPPPQGFKQEYHDPLYEHGVPGMPGPPAHGFQSPM

Application

Monoclonal Anti-ETV5 antibody produced in mouse has been used in western blotting (1:1000).

Biochem/physiol Actions

E-twenty-six (Ets) variant gene 5 (ETV5) gene is responsible for the survival, proliferation, and self-renewal of spermatogonial stem cells (SSCs). ETV5 protein plays a role in mediating glial cell line-derived neurotrophic factor (GDNF) signaling and induces several genes required for regulating SSC fate. It is involved in limb development and regulates epithelial-mesenchymal transition in several cancer cells. ETV5 protein binds to the conserved GGAA/T motif and regulates gene expression.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erik Melén et al.
The Journal of allergy and clinical immunology, 126(3), 631-637 (2010-09-08)
Epidemiologic studies consistently show associations between asthma and obesity. Shared genetics might account for this association. We sought to identify genetic variants associated with both asthma and obesity. On the basis of a literature search, we identified genes from (1)
Lee Huang et al.
Cancer research, 81(8), 2071-2085 (2021-02-03)
The failure of once promising target-specific therapeutic strategies often arises from redundancies in gene expression pathways. Even with new melanoma treatments, many patients are not responsive or develop resistance, leading to disease progression in terms of growth and metastasis. We
Xin Wu et al.
Biology of reproduction, 85(6), 1114-1123 (2011-08-06)
Insight regarding mechanisms controlling gene expression in the spermatogonial stem cell (SSC) will improve our understanding of the processes regulating spermatogenesis and aid in treating problems associated with male infertility. In the present study, we explored the global gene expression
Duy Pham et al.
The Journal of allergy and clinical immunology, 134(1), 204-214 (2014-02-04)
The differentiation of TH17 cells, which promote pulmonary inflammation, requires the cooperation of a network of transcription factors. We sought to define the role of Etv5, an Ets-family transcription factor, in TH17 cell development and function. TH17 development was examined
Severa Bunda et al.
Nature communications, 10(1), 661-661 (2019-02-10)
Capicua (CIC) is a transcriptional repressor that counteracts activation of genes downstream of receptor tyrosine kinase (RTK)/Ras/ERK signaling. It is well-established that tumorigenesis, especially in glioblastoma (GBM), is attributed to hyperactive RTK/Ras/ERK signaling. While CIC is mutated in other tumors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service