Skip to Content
Merck
All Photos(3)

Key Documents

V4139

Sigma-Aldrich

Anti-Vinculin Antibody

enhanced validation

rabbit polyclonal

Synonym(s):

Vinculin Antibody - Anti-Vinculin antibody produced in rabbit, Vinculin Antibody Sigma

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

Product Name

Anti-Vinculin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~116 kDa

species reactivity

rat, human, mouse

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

indirect immunofluorescence: 15-30 μg/mL using acetone-fixed frozen sections of rat or human heart
western blot: 2.5-5 μg/mL using whole extracts of cultured mouse NIH3T3 cells and human hepatoma HepG2 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VCL(7414)
mouse ... Vcl(22330)
rat ... Vcl(305679)

General description

In muscle, vinculin is localized in the fascia adherens of the intercalated disk (cardiac muscle), myotendinous junctions (skeletalmuscle), neuromuscular junctions, and the membrane associated dense bodies of smooth muscle. In many cell types undergoing viral transformation, vinculin becomes redistributed to rosettes or podosomes.
Vinculin (VCL) is a ubiquitously expressed cytoskeletal protein, encoded by the gene mapped to human chromosome 10q11.2-qter. VCL is a vital element of adherens type cell junctions. Vinculin structure contains a globular head connected to a tail domain by a short proline-rich sequence, and an intramolecular interaction between the head and tail hides the various ligand-binding sites in the protein.

Immunogen

synthetic peptide corresponding to amino acid residues 691-705 of human vinculin with N-terminal added cysteine, conjugated to KLH. This sequence is also present in metavinculin. The corresponding sequence is identical is mouse, rat, and pig and differs by one amino acid in chicken.

Application

Anti-Vinculin antibody produced in rabbit has been used in immunoprecipitation.
Anti-vinculin antibody produced in rabbit has been used in following studies:
  • immunofluorescence
  • immuno-histology
  • western blotting

Biochem/physiol Actions

Vinculin (VCL) is a cytoskeletal protein that supports cell-to-cell and cell to matrix junctions. It is also essentially involved in normal embryonic development and cardiac function, due to its role in controlling adhesion and motility, as well as cell spreading. In mature focal adhesions (FAs), VCL acts as an important constituent of the “molecular clutch” that induces the transmission of force from cytoplasmic F-actin to membrane-bound integrins. Aberration or mutation of VCL leads to cardiomyopathy, which is associated with hypertension. Experimental studies reveal that, VCL might be used as a biomarker of aging or heart failure. Vinculin associates with FAs and assists in several cellular functions, such as stabilizing and strengthening FAs and promoting their maturation, extracellular matrix (ECM) mechanosensing, regulating actin cytoskeletal dynamics and signaling pathways involved in apoptosis.
Vinculin also binds α-catenin as well as the vasodilator stimulated phosphoprotein (VASP) in focal contact assembly. It transiently binds the actin-related protein-2/3 (ARP2/3) complex without colocalization in focal adhesions.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher J Walker et al.
Oncotarget, 8(40), 68758-68768 (2017-10-06)
Extensive genomic profiling for endometrioid endometrial carcinoma (EEC) has pointed to genes and pathways important in uterine development as critical mediators of endometrial tumorigenesis. SOX17 is a developmental transcription factor necessary for proper endoderm formation that has been implicated as
Sex-specific association of rs4746172 of VCL gene with hypertension in two Han populations from Southern China.
Yu Q
Scientific Reports (2015)
How vinculin regulates force transmission
Dumbauld DW, et al.
Proceedings of the National Academy of Sciences of the USA, 110(24), 9788-9793 (2013)
M Itoh et al.
The Journal of cell biology, 115(5), 1449-1462 (1991-12-01)
Recently we developed an isolation procedure for the cell-to-cell adherens junctions (AJ; cadherin-based junctions) from rat liver (Tsukita, Sh. and Sa. Tsukita. 1989. J. Cell Biol. 108:31-41). In this study, using the isolated AJ, we have obtained two mAbs specific
The differential effects of aligned electrospun PHBHHx fibers on adipogenic and osteogenic potential of MSCs through the regulation of PPARg signaling
Wang Y
Biomaterials, 33, 485-493 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service