Skip to Content
Merck
All Photos(1)

Documents

V3640

Sigma-Aldrich

Valpromide

≥97% (NMR)

Synonym(s):

2-Propylvaleramide, 2-propyl-pentanamide, Depamid, Depamide, Di-n-propylacetamide, Dipropylacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H17NO
CAS Number:
Molecular Weight:
143.23
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥97% (NMR)

form

powder

color

white to off-white

solubility

DMSO: >10 mg/mL

originator

Sanofi Aventis

storage temp.

room temp

SMILES string

CCCC(CCC)C(N)=O

InChI

1S/C8H17NO/c1-3-5-7(6-4-2)8(9)10/h7H,3-6H2,1-2H3,(H2,9,10)

InChI key

OMOMUFTZPTXCHP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Valpromide has been used to pretreat NIH/3T3 cells to test its effect on cytomegalovirus (CMV) viral replication. It has also been used to test its antiviral functionality in herpes simplex virus type 1 (HSV-1) infected human oligodendroglioma (HOG) cells. It may be used to test its effect on apoptosis induction in astrocytes.

Biochem/physiol Actions

Valpromide (VPD) is a derivative of valproic acid (VPA) and is used as an antiepileptic drug. It is hydrolyzed quickly to VPA in vivo, but has intrinsic anticonvulsant activity.
Valpromide possesses antipsychotic property. It lacks the toxic and teratogenic effects of valproic acid. It also lacks the histone deacetylase (HDAC) inhibitory activity of valproic acid.

Features and Benefits

This compound was developed by Sanofi Aventis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J A Shimshoni et al.
Molecular pharmacology, 71(3), 884-892 (2006-12-15)
Valproic acid (VPA) is an effective antiepileptic drug with an additional activity for the treatment of bipolar disorder. It has been assumed that both activities arise from a common target. At the molecular level, VPA targets a number of distinct
Karine Cambon et al.
Molecular therapy. Methods & clinical development, 5, 259-276 (2017-06-13)
Huntington's disease (HD) is an autosomal dominant neurodegenerative disorder resulting from a polyglutamine expansion in the huntingtin (HTT) protein. There is currently no cure for this disease, but recent studies suggest that RNAi to downregulate the expression of both normal
Valproic acid induces apoptosis in differentiating hippocampal neurons by the release of tumor necrosis factor-alpha from activated astrocytes
Wang C, et al.
Neuroscience Letters, 497(2), 122-127 (2011)
Cheng-Wei Li et al.
PloS one, 13(8), e0202537-e0202537 (2018-08-23)
Epstein-Barr virus (EBV), also known as human herpesvirus 4, is prevalent in all human populations. EBV mainly infects human B lymphocytes and epithelial cells, and is therefore associated with their various malignancies. To unravel the cellular mechanisms during the infection
Akinobu Okada et al.
Birth defects research. Part B, Developmental and reproductive toxicology, 71(1), 47-53 (2004-03-03)
The antiepileptic drug valproic acid (VPA) is well known to cause neural tube and skeletal defects in both humans and animals. The amidic VPA analogues valpromide (VPD) and valnoctamide (VCD) have much lower teratogenicity than VPA inducing exencephaly in mice.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service