Skip to Content
Merck
All Photos(5)

Key Documents

U5625

Sigma-Aldrich

Uridine 5′-diphosphoglucuronic acid ammonium salt

98-100%

Synonym(s):

UDP-GlcA, UDPGA, Uridine-diphosphate-glucuronic acid ammonium salt, Uridine[5′]diphospho[1]-α-D-glucopyranosuronic acid ammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22N2O18P2 · xNH3
CAS Number:
Molecular Weight:
580.29 (free acid basis)
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

Saccharomyces cerevisiae
enzyme from bovine liver (catalase)
enzyme from rabbit muscle (LDH)

Assay

98-100%

form

powder

storage temp.

−20°C

SMILES string

N.O[C@@H]1[C@@H](O)[C@H](O[C@@H]([C@H]1O)C(O)=O)OP(O)(=O)OP(O)(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N3C=CC(=O)NC3=O

InChI

1S/C15H22N2O18P2.H3N/c18-5-1-2-17(15(26)16-5)12-9(22)6(19)4(32-12)3-31-36(27,28)35-37(29,30)34-14-10(23)7(20)8(21)11(33-14)13(24)25;/h1-2,4,6-12,14,19-23H,3H2,(H,24,25)(H,27,28)(H,29,30)(H,16,18,26);1H3/t4-,6-,7+,8+,9-,10-,11+,12-,14-;/m1./s1

InChI key

WMWKTCPGFOEPBD-YGIWDPDDSA-N

General description

Uridine 5′-diphosphoglucuronic acid (UDPGA) is a cofactor, synthesized from UDPglucose and nicotinamide adenine dinucleotide (NAD+) intracellularly by the catalytic activity of UDPglucose dehydrogenase. The concentration of UDPGA in a number of liver preparations ranges from 0.3-0.5 mM.

Application

Uridine 5′-diphosphoglucuronic acid ammonium salt has been used in the initiation of UDP-glucuronosyl transferase (UDGPT) assay towards 14C-testosterone in the liver S9 protein. It is suitable for use in UDGPT assay.

Biochem/physiol Actions

Uridine 5′-diphosphoglucuronic acid (UDGPA) is the glucuronic acid donor for the conjugation of bilirubin in the endoplasmic recticulum.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shupeng Yang et al.
Journal of agricultural and food chemistry, 65(33), 7217-7227 (2017-07-25)
After being incubated with animal and human liver microsomes, metabolites of phase I and II were investigated. A comparison was performed by ultrahigh performance liquid chromatography-quadrupole/time-of-flight coupled to mass spectrometry (UHPLC-Q/TOF). Consequently, a total of four phase I metabolites and
Jimmy Flarakos et al.
Xenobiotica; the fate of foreign compounds in biological systems, 47(8), 682-696 (2016-08-09)
1. Absorption, distribution, metabolism, transport and elimination properties of omadacycline, an aminomethylcycline antibiotic, were investigated in vitro and in a study in healthy male subjects. 2. Omadacycline was metabolically stable in human liver microsomes and hepatocytes and did not inhibit or induce
Molly Lin et al.
Xenobiotica; the fate of foreign compounds in biological systems, 48(10), 1021-1027 (2017-08-29)
1. Glucuronidation of amines has been shown to exhibit large species differences, where the activity is typically more pronounced in human than in many preclinical species such as rat, mouse, dog and monkey. The purpose of this work was to
Seasonal testosterone UDP-glucuronosyltransferase activity and biliary steroids in Eurasian perch: Response to leachate exposure
Linderoth M, et al.
Ecotoxicology and Environmental Safety, 68(1), 49-56 (2007)
Justine Badée et al.
Drug metabolism and disposition: the biological fate of chemicals, 47(2), 124-134 (2018-11-28)
UDP-glucuronosyltransferase (UGT)-mediated metabolism is possibly the most important conjugation reaction for marketed drugs. However, there are currently no generally accepted standard incubation conditions for UGT microsomal assays, and substantial differences in experimental design and methodology between laboratories hinder cross-study comparison

Articles

Explore tools for glycosyltransferase synthesis and modification of glycans, such as glycosyltransferases and nucleotide sugar donors.

Explore tools for glycosyltransferase synthesis and modification of glycans, such as glycosyltransferases and nucleotide sugar donors.

LC-MS/MS method quantifies similar polar nucleotide activated sugars using Supel™ Carbon LC column for simultaneous analysis.

LC-MS/MS method quantifies similar polar nucleotide activated sugars using Supel™ Carbon LC column for simultaneous analysis.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service