Skip to Content
Merck
All Photos(2)

Key Documents

T8390

Sigma-Aldrich

Testosterone 3-(O-carboxymethyl)oxime

Synonym(s):

17β-Hydroxy-4-androsten-3-one 3-(O-carboxymethyl)oxime, 4-Androsten-17β-ol-3-one 3-(O-carboxymethyl)oxime

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H31NO4
CAS Number:
Molecular Weight:
361.48
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

sterility

non-sterile

Quality Level

Assay

≥98.00% (TLC)

form

powder

drug control

Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada

solubility

ethanol: soluble 49-51 mg/mL, clear, colorless to faintly yellow

shipped in

ambient

storage temp.

room temp

SMILES string

[H][C@@]12CCC3=CC(\CC[C@]3(C)[C@@]1([H])CC[C@]4(C)[C@@H](O)CC[C@@]24[H])=N\OCC(O)=O

InChI

1S/C21H31NO4/c1-20-9-7-14(22-26-12-19(24)25)11-13(20)3-4-15-16-5-6-18(23)21(16,2)10-8-17(15)20/h11,15-18,23H,3-10,12H2,1-2H3,(H,24,25)/b22-14+/t15-,16-,17-,18-,20-,21-/m0/s1

InChI key

VDYLVWGBLQNNAW-UVTKPVMCSA-N

Application

Testosterone 3-(O-carboxymethyl)oxime has been used for the hormonal analysis of sexual dimorphism in the electric fish, Apteronotus. This testosterone derivative has also been used for developing environmental biosensors for the detection of testosterone in water samples.
Testosterone 3-(O-carboxymethyl)oxime has been used to prepare horseradish peroxidase (HRP)-labeled testosterone to measure plasma testosterone concentration by enzyme immunoassay.

Biochem/physiol Actions

Androgens direct the development of male reproductive system and secondary sexual characteristics. Testosterone is an androgen that is secreted by the testis. This hormone is converted to dihydrotestosterone (DHT) by 5α reductase in the target tissues. Both DHT and testosterone require androgen receptor, a ligand-dependent nuclear transcription factor to mediate their biological functions.
Androgens direct the development of the male phenotype during embryogenesis and at puberty. Testosterone is an androgen that is secreted by the testis. This hormone is converted to dihydrotestosterone in the target tissues where it may mediate several biological functions.

Preparation Note

Testosterone 3-(O-carboxymethyl)oxime is soluble in ethanol at 49-51 mg/ml and yields a clear, colorless to faint yellow solution.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Testosterone and financial risk preferences
Apicella CL, et al.
Evolution, 29(6), 384-390 (2008)
SB223412, a neurokinin-3 receptor-selective antagonist, suppresses testosterone secretion in male guinea pigs
Nakamura S, et al.
Theriogenology, 102, 183-189 (2017)
Androgen receptor structure, function and biology: from bench to bedside
Davey RA and Grossmann M
The Clinical Biochemist. Reviews / Australian Association of Clinical Biochemists, 37(1), 3-3 (2016)
K D Dunlap et al.
Journal of comparative physiology. A, Sensory, neural, and behavioral physiology, 183(1), 77-86 (1998-08-06)
Gymnotiform electric fish emit an electric organ discharge that, in several species, is sexually dimorphic and functions in gender recognition. In addition, some species produce frequency modulations of the electric organ discharge, known as chirps, that are displayed during aggression
Jens Tschmelak et al.
Talanta, 69(2), 343-350 (2008-10-31)
Nowadays, little technology exists that can monitor various water sources quickly and at a reasonable cost. The ultra-sensitive, fully automated and robust biosensor River Analyser (RIANA) is capable of detecting multiple organic targets rapidly and simultaneously at a heterogeneous assay

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service