Skip to Content
Merck
All Photos(5)

Key Documents

T7816

Sigma-Aldrich

Monoclonal Anti-β-Tubulin I antibody produced in mouse

clone SAP.4G5, ascites fluid

Synonym(s):

Anti-CDCBM6, Anti-CSCSC1, Anti-M40, Anti-OK/SW-cl.56, Anti-TUBB1, Anti-TUBB5

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

SAP.4G5, monoclonal

mol wt

antigen ~55 kDa

species reactivity

Xenopus, chicken, human, hamster, canine, mouse, bovine, rat

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
microarray: suitable
western blot: 1:20,000 using cultured chicken fibroblasts

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TUBB(203068)
mouse ... Tubb4(22153)

General description

Tubulin a 100kD protein is present in all cells as a heterodimer of two similar polypeptides α and β which assemble to form microtubules. Proper organization of microtubules is essential for several cellular functions like mitosis, meiosis, some forms of organellar movement, intracellular transport, flagellar movement and cytoskeletal functions. Monoclonal anti-β-tubulin I antibody can be used in ELISA to localize β tubulin isotype I and can be used in immunoblotting to localize cultured human and animal cells. Mouse anti-β-tubulin I antibody reacts specifically with an epitope found in the c-terminal sequence of β tubulin of human, rat, dog, bovine, hamster, mouse, chicken and xenopus

Specificity

Recognizes an epitope located in the C-terminal sequence of β-tubulin isotype I. No reactivity with other tubulin isotypes is observed.

Immunogen

synthetic peptide corresponding to the C-terminal sequence of β-tubulin isotype I, conjugated to BSA.

Application

Monoclonal anti-β-tubulin I antibody (diluted 1: 4,000) can be used as a primary antibody in immunoprecipitation and immunoblotting. It can also be used in western blot analysis and retroviral infection.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Richard G James et al.
PloS one, 7(12), e50457-e50457 (2012-12-12)
The Wnt/ß-catenin signaling pathway controls important cellular events during development and often contributes to disease when dysregulated. Using high throughput screening we have identified a new small molecule inhibitor of Wnt/ß-catenin signaling, WIKI4. WIKI4 inhibits expression of ß-catenin target genes
Yosef Buganim et al.
Cancer research, 66(22), 10750-10759 (2006-11-17)
Mutations in p53 are ubiquitous in human tumors. Some p53 mutations not only result in loss of wild-type (WT) activity but also grant additional functions, termed "gain of function." In this study, we explore how the status of p53 affects
Ohad Tarcic et al.
Cell reports, 14(6), 1462-1476 (2016-02-09)
Factors linking inflammation and cancer are of great interest. We now report that the chromatin-targeting E3 ubiquitin ligase RNF20/RNF40, driving histone H2B monoubiquitylation (H2Bub1), modulates inflammation and inflammation-associated cancer in mice and humans. Downregulation of RNF20 and H2Bub1 favors recruitment
M C Roach et al.
Cell motility and the cytoskeleton, 39(4), 273-285 (1998-05-15)
Tubulin, the subunit protein of microtubules, is an alpha/beta heterodimer. In many organisms, both alpha and beta consist of various isotypes. Although the isotypes differ in their tissue distributions, the question of whether the isotypes perform different functions in vivo
Richard G James et al.
The Journal of biological chemistry, 288(48), 34658-34670 (2013-10-12)
Advances in phosphoproteomics have made it possible to monitor changes in protein phosphorylation that occur at different steps in signal transduction and have aided the identification of new pathway components. In the present study, we applied this technology to advance

Articles

Microtubules of the eukaryotic cytoskeleton are composed of a heterodimer of α- and β-tubulin. In addition to α-and β-tubulin, several other tubulins have been identified, bringing the number of distinct tubulin classes to seven.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service