Skip to Content
Merck
All Photos(2)

Key Documents

T6513

Sigma-Aldrich

Glyceryl trilinolenate

≥97% (TLC), liquid

Synonym(s):

TG(18:3(9Z,12Z,15Z)/18:3(9Z,12Z,15Z)/18:3(9Z,12Z,15Z)), 1,2,3-Tri-(cis,cis,cis-9,12,15-octadecatrienoyl)glycerol, 1,2,3-Trilinolenoylglycerol, Glycerol trilinolenate, Trilinolenin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C57H92O6
CAS Number:
Molecular Weight:
873.34
Beilstein:
1718706
EC Number:
MDL number:
UNSPSC Code:
41141833
PubChem Substance ID:
NACRES:
NA.25

biological source

Linum usitatissimum oil

Assay

≥97% (TLC)

form

liquid

refractive index

n20/D 1.489

density

0.94 g/mL at 20 °C (lit.)

functional group

ester

lipid type

neutral glycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC\C=C/C\C=C/C\C=C/CCCCCCCC(=O)OCC(COC(=O)CCCCCCC\C=C/C\C=C/C\C=C/CC)OC(=O)CCCCCCC\C=C/C\C=C/C\C=C/CC

InChI

1S/C57H92O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-49-55(58)61-52-54(63-57(60)51-48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)53-62-56(59)50-47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h7-12,16-21,25-30,54H,4-6,13-15,22-24,31-53H2,1-3H3/b10-7-,11-8-,12-9-,19-16-,20-17-,21-18-,28-25-,29-26-,30-27-

InChI key

UBEIMDKGOYBUKT-FLIQGJDUSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Quantitation of furan and methylfuran formed in different precursor systems by proton transfer reaction mass spectrometry.: The research quantifies furan and methylfuran production from various precursors using advanced mass spectrometry techniques. These insights are important for food safety and understanding the chemical reactions during food processing (Märk et al., 2006).

Packaging

Sealed ampule.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kanika Mitra et al.
Journal of food science, 77(1), C39-C45 (2011-11-30)
Alpha-linolenic acid (ALA) enriched structured lipid (SL) was produced by lipase-catalyzed interesterification from perilla oil (PO) and corn oil (CO). The effects of different reaction conditions (substrate molar ratio [PO/CO 1:1 to 1:3], reaction time [0 to 24 h], and
Glen S Patten et al.
Digestive diseases and sciences, 54(3), 511-521 (2008-07-12)
The aim of this study was to design food grade matrices to deliver microencapsulated fish oil to the large bowel of the rat where the potential exists to retard inflammation and cancer development. Digestion in simulated gastric fluid and intestinal
Qin Guo et al.
Journal of the science of food and agriculture, 97(14), 4697-4703 (2017-04-04)
The polarities of linolenic acid isomers are very similar, and only a few studies to date have attempted to separate α-linolenic acid (ALA) isomers completely. The aim of this study was to fill this gap by developing and validating an
Beatriz Lázaro et al.
PloS one, 12(4), e0176520-e0176520 (2017-04-28)
Triglycerides (TAGs), the major storage molecules of metabolic energy and source of fatty acids, are produced as single cell oil by some oleogenic microorganisms. However, these microorganisms require strict culture conditions, show low carbon source flexibilities, lack efficient genetic modification
Karen M MacDougall et al.
Analytical and bioanalytical chemistry, 401(8), 2609-2616 (2011-09-15)
Biofuels from photosynthetic microalgae are quickly gaining interest as a viable carbon-neutral energy source. Typically, characterization of algal feedstock involves breaking down triacylglycerols (TAG) and other intact lipids, followed by derivatization of the fatty acids to fatty acid methyl esters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service