Skip to Content
Merck
All Photos(2)

Key Documents

T5030

Sigma-Aldrich

Tris Buffered Saline

tablet

Synonym(s):

TBS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161700
NACRES:
NA.25

Product Name

Tris Buffered Saline, tablet

form

tablet

Quality Level

application(s)

diagnostic assay manufacturing

Looking for similar products? Visit Product Comparison Guide

Application

Tris Buffered Saline Tablets are suitable for use in immunoassay procedures.
It may be used in the following studies:
  • Immunogold-silver staining (IGSS) method to investigate the immunoglobulins in reactive human tonsil.
  • To study the aggregation of purified samples of α-synuclein.
  • As buffer in Western Blot analysis.
  • Time resolved fluoroimmunoassay.
  • As buffer in light microscopic immunohistochemistry.

Reconstitution

One tablet dissolved in 15 mL of deionized water yields 0.05 M Tris and 0.15 M sodium chloride, pH 7.6, at 25 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J E Meredith et al.
Molecular biology of the cell, 4(9), 953-961 (1993-09-01)
Programmed cell death (PCD) or apoptosis is a naturally occurring cell suicide pathway induced in a variety of cell types. In many cases, PCD is induced by the withdrawal of specific hormones or growth factors that function as survival factors.
Katarzyna Herman et al.
ACS chemical biology, 14(7), 1611-1618 (2019-07-10)
Lectin-carbohydrate interactions can be exploited in ultrasensitive biochemical recognition or medical diagnosis. For this purpose, besides the high specificity of the interactions, an appropriate methodology for their quantitative and detailed characterization is demanded. In this work, we determine the unbinding
C S Holgate et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 31(7), 938-944 (1983-07-01)
A new method for demonstrating antigens in paraffin sections of formol sublimate-fixed tissue is described that utilizes an "indirect" immunohistological technique employing immunoglobulin adsorbed to colloidal gold as the secondary antiserum. The gold particles introduced to antigenic sites are revealed
G Pagès et al.
Proceedings of the National Academy of Sciences of the United States of America, 90(18), 8319-8323 (1993-09-15)
The mitogen-activated protein kinases (MAP kinases) p42mapk and p44mapk are serine/threonine kinases rapidly activated in cells stimulated with various extracellular signals by dual phosphorylation of tyrosine and threonine residues. They are thought to play a pivotal role in integrating and
A J Maniotis et al.
The American journal of pathology, 155(3), 739-752 (1999-09-17)
Tissue sections from aggressive human intraocular (uveal) and metastatic cutaneous melanomas generally lack evidence of significant necrosis and contain patterned networks of interconnected loops of extracellular matrix. The matrix that forms these loops or networks may be solid or hollow.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service